Skip to Content
Merck
All Photos(1)

Key Documents

233188

Sigma-Aldrich

4′-(Trifluoromethyl)acetophenone

98%

Synonym(s):

4-Acetylbenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4COCH3
CAS Number:
Molecular Weight:
188.15
Beilstein:
1870425
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

bp

79-80 °C/8 mmHg (lit.)

mp

30-33 °C (lit.)

functional group

fluoro
ketone

SMILES string

CC(=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C9H7F3O/c1-6(13)7-2-4-8(5-3-7)9(10,11)12/h2-5H,1H3

InChI key

HHAISVSEJFEWBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The enantioselective addition of dialkylzinc to 4′-(trifluoromethyl)acetophenone mediated by 1,2-bis(hydroxycamphorsulfonamido)cyclohexenes in the presence of titanium tetraisopropoxide has been investigated[1]. Phosphorescence emission spectra of 4′-(trifluoromethyl)acetophenone has been studied using pulsed source phosphorimetry[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pulsed source, time resolved phosphorimetry determination of phosphorescence lifetimes.
Harbaugh KF, et al.
Analytical Chemistry, 45(2), 381-382 (1973)
Highly enantioselective addition of dialkylzinc reagents to ketones promoted by titanium tetraisopropoxide.
Yus M, et al.
Tetrahedron Asymmetry, 13(21), 2291-2293 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service