Skip to Content
Merck
All Photos(1)

Key Documents

390380

Sigma-Aldrich

4-Acetamido-TEMPO, free radical

97%

Synonym(s):

4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21N2O2
CAS Number:
Molecular Weight:
213.30
Beilstein:
3546225
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

143-145 °C (lit.)

SMILES string

CC(=O)NC1CC(C)(C)N([O])C(C)(C)C1

InChI

1S/C11H21N2O2/c1-8(14)12-9-6-10(2,3)13(15)11(4,5)7-9/h9H,6-7H2,1-5H3,(H,12,14)

InChI key

UXBLSWOMIHTQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxidizes alcohols to carbonyl compounds in the presence of TsOH.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erika Watanabe et al.
Carbohydrate polymers, 100, 74-79 (2013-11-06)
Pure (1→3)-β-polyglucuronic acid sodium salt was prepared from curdlan by oxidation with 4-acetamido-TEMPO/NaClO/NaClO₂ in water at pH 4.7 and 35°C. The oxidation conditions, including the reaction time and amounts of reagents added, were optimized for the preparation of (1→3)-β-polyglucuronic acids
Oxidation of vic-Diols to α-dicarbonyl compounds using the oxoammonium salt derived from 4-acetamido-TEMPO and p-toluenesulfonic acid.
Banwell MG, et al.
The Journal of Organic Chemistry, 59(21), 6338-6343 (1994)
Lenka Vítková et al.
Polymers, 11(9) (2019-09-22)
In the current study, we present methods of sodium hyaluronate, also denoted as hyaluronan (HA), nanofiber fabrication using a direct-current (DC) electric field. HA was spun in combination with poly(vinyl alcohol) (PVA) and polyethylene oxide (PEO) and as a pure
The Journal of Organic Chemistry, 56, 6110-6110 (1991)
Iron Chloride/4-Acetamido-TEMPO/Sodium Nitrite-Catalyzed Aerobic Oxidation of Primary Alcohols to the Aldehydes.
Yin W, et al.
Advanced Synthesis & Catalysis, 352(1, 113-118 (2010)

Related Content

he Stahl Lab focuses on the development of catalysts and catalytic reactions for selective oxidation of organic molecules, with particular emphasis on aerobic oxidation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service