Skip to Content
Merck
All Photos(1)

Key Documents

382000

Sigma-Aldrich

4-Carboxy-TEMPO, free radical

97%

Synonym(s):

4-Carboxy-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-Carboxy-2,2,6,6-tetramethylpiperidinyloxy, free radical, 4-Carboxy-TEMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18NO3
CAS Number:
Molecular Weight:
200.25
Beilstein:
3949026
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

185-189 °C (lit.)

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

CC1(C)CC(CC(C)(C)N1[O])C(O)=O

InChI

1S/C10H18NO3/c1-9(2)5-7(8(12)13)6-10(3,4)11(9)14/h7H,5-6H2,1-4H3,(H,12,13)

InChI key

CYQGCJQJIOARKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Carboxy-TEMPO is a 4-substituted 2,2,6,6-tetramethylpiperidyl-1-oxy (TEMPO) derivative. 4-carboxy-TEMPO immobilized with 3-aminopropylsilane on the mesoporous silica surface comprises heterogeneous catalytic system employed for various oxidation reactions.

Application

4-Carboxy-TEMPO may be employed as catalyst for the oxidation of regenerated cellulose (viscose rayon) to cellouronic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuyuki Hosokawa et al.
Free radical biology & medicine, 37(7), 946-952 (2004-09-01)
Three water-soluble carboxy nitroxide antioxidants, 5-carboxy-1,1,3,3-tetramethylisoindolin-2-yloxyl, 4-carboxy-2,2,6,6-tetramethylpiperidin-1-yloxyl, and 3-carboxy-2,2,5,5-tetramethylpyrrolidin-1-yloxyl, show significant impact on the postirradiation survival rates of ataxia telangiectasia (A-T) cells compared to normal cells, an assay which represents a model for understanding the impact of ROS damage on
X Yang et al.
Biophysical journal, 71(3), 1587-1595 (1996-09-01)
Electron paramagnetic resonance spectroscopy and gel permeation chromatography were employed to study the molecular diffusion of a number of small nitroxide spin probes (approximately 7-9 A diameter) into the central cavity of the iron-storage protein ferritin. Charge and polarity of
Comparison of two MCM-41 grafted TEMPO catalysts in selective alcohol oxidation.
Brunel D, et al.
Applied Catalysis A: General, 213(1), 73-82 (2001)
Nitroxide-mediated oxidation of cellulose using TEMPO derivatives: HPSEC and NMR analyses of the oxidized products.
Shibata I and Isogai A.
Cellulose, 10(4), 335-341 (2003)
J L Lamarque et al.
European journal of radiology, 6(1), 48-52 (1986-02-01)
The meglumine salt of TCA, a piperidinyl nitroxide stable free radical derivative, was tested as an NMR contrast enhancer. This report of its use in animals indicates its usefulness in the enhancement of signal intensity of the kidney and bladder.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service