Skip to Content
Merck
All Photos(2)

Documents

371904

Sigma-Aldrich

1,3-Divinyltetramethyldisiloxane

97%

Synonym(s):

1,1,3,3-Tetramethyl-1,3-divinyldisiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHSi(CH3)2]2O
CAS Number:
Molecular Weight:
186.40
Beilstein:
1762585
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

139 °C (lit.)

mp

−99 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(O[Si](C)(C)C=C)C=C

InChI

1S/C8H18OSi2/c1-7-10(3,4)9-11(5,6)8-2/h7-8H,1-2H2,3-6H3

InChI key

BITPLIXHRASDQB-UHFFFAOYSA-N

Application


  • Synthesis and Studies on the Effect of Phenyl Side–Chain Content on Refractive Index of Polysiloxane Resin: This study used 1,3-divinyltetramethyldisiloxane as a terminating agent to modify polysiloxane resin properties (Ramli et al., 2015).

  • Undecenoic Acid-Based Polydimethylsiloxanes Obtained by Hydrosilylation and Hydrothiolation Reactions: This paper explored the use of 1,3-divinyltetramethyldisiloxane as a blocking agent in the synthesis of polydimethylsiloxanes (Milenin et al., 2020).

  • Efficient Improvement in Electrochemical Properties of High-Voltage Li-rich Mn-Based Layered Oxide Cathode by Addition of 1,3-divinyltetramethyldisiloxane: The additive 1,3-divinyltetramethyldisiloxane was used to enhance electrochemical properties of Li-rich layered oxide cathode (Huang et al., 2023).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.1 °F - closed cup

Flash Point(C)

21.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Sivaranjini et al.
Scientific reports, 8(1), 8891-8891 (2018-06-13)
A simple and effective approach for vertical alignment of liquid crystals (LCs) over a functionalized transparent flexible substrate is described. Surface characterization of this commercially available plastic substrate through X-ray photoelectron spectroscopy (XPS) and attenuated total reflection infrared spectroscopy (ATR-IR)
Seungmin Shin et al.
Nanotechnology, 27(47), 475301-475301 (2016-10-27)
Novel polyhedral oligomeric silsesquioxane (POSS) resists, which are based on a new photo-crosslinking system via Wolff rearrangement, are developed as ideal replica mold materials for ultraviolet-nanoimprint lithography. These POSS resist materials are synthesized by incorporating diazoketo and hydroxyl groups into
Charles A Ault et al.
Macromolecular rapid communications, 42(5), e2000692-e2000692 (2021-01-28)
Effects of molecular weight of methylphenyl-containing vinylsiloxy-functionalized terpolysiloxanes on their UV-activated crosslinking by hydrosilylation at room temperature in air, shelf life stability of "all-in-one" pastes prepared from them for additive manufacturing, and mechanical properties of the resulting crosslinked elastomers, are
Katharina Thum et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(7), 2513-2522 (2020-11-17)
The first intermolecular early main group metal-alkene complexes were isolated. This was enabled by using highly Lewis acidic Mg centers in the Lewis base-free cations (Me BDI)Mg+ and (tBu BDI)Mg+ with B(C6 F5 )4 - counterions (Me BDI=CH[C(CH3 )N(DIPP)]2
Ilya Borisov et al.
Polymers, 12(6) (2020-05-30)
For the first time, the effect of the side-chain in polyalkylmethylsiloxane towards pervaporative removal of methyl tert-butyl ether (MTBE) from water was studied. The noticeable enhancement of separation factor during the pervaporation of 1 wt.% MTBE solution in water through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service