Skip to Content
Merck
All Photos(1)

Documents

235695

Sigma-Aldrich

Octamethylcyclotetrasiloxane

98%

Synonym(s):

2,2,4,4,6,6,8,8-Octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)2O-]4
CAS Number:
Molecular Weight:
296.62
Beilstein:
1787074
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

1 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

175-176 °C (lit.)

mp

17-18 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

InChI

1S/C8H24O4Si4/c1-13(2)9-14(3,4)11-16(7,8)12-15(5,6)10-13/h1-8H3

InChI key

HMMGMWAXVFQUOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octamethylcyclotetrasiloxane (D4) is a volatile methylsiloxane (VMS) with a relatively low molecular weight. This silicone fluid has a silicon-oxygen bond in a cyclic arrangement and methyl groups are attached to the silicon atom. D4 can be used in the manufacture of silicone based polymers for use in medical devices and personal care products.

Application

D4 can be polymerized through ring opening mechanism with a rare earth solid super acid as a catalyst for potential usage in the synthesis of vinyl coated polydimethylsiloxane (PDMS). It may also be used as a probe liquid for nuclear magnetic resonance cryoporometry (NMRC) to analyze the pore size distribution (PSD) of oil-bearing tight sandstone. Chemical vapor deposition (CVD) of D4 may be utilized for the preparation of siloxane based thin films.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Watermann et al.
Metabolites, 11(1) (2021-01-13)
1H NMR spectroscopy, in combination with chemometric methods, was used to analyze the methanol/acetonitrile (1:1) extract of walnut (Juglans Regia L.) regarding the geographical origin of 128 authentic samples from different countries (France, Germany, China) and harvest years (2016-2019). Due
Hot-filament chemical vapor deposition of organosilicon thin films from hexamethylcyclotrisiloxane and octamethylcyclotetrasiloxane.
Lewis HGP
Journal of the Electrochemical Society, 148(12), F212-F220 (2001)
Applying octamethylcyclotetrasiloxane as a probe liquid for characterizing the pore size distribution of oil-bearing tight sandstones by nuclear magnetic resonance cryoporometry.
Liu Biao, et al.
Marine and Petroleum Geology, 88(2), 814-825 (2017)
Photocatalytic oxidation of octamethylcyclotetrasiloxane (D4): Towards a better understanding of the impact of volatile methyl siloxanes on photocatalytic systems.
Lamaa L, et al.
Applied Catalysis. B, Environmental, 156, 438-446 (2014)
Ramesh Sarangapani et al.
Toxicological sciences : an official journal of the Society of Toxicology, 67(2), 159-172 (2002-05-16)
Inhalation of octamethylcyclotetrasiloxane (D4) induces CYP2B1/2 protein and causes liver enlargement. We have developed a pharmacodynamic (PD) extension to a physiologically based pharmacokinetic (PBPK) model to characterize these dose-response behaviors. The PD model simulates interactions of D4 with a putative

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service