Skip to Content
Merck
All Photos(2)

Key Documents

367044

Sigma-Aldrich

(3S)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione

98%

Synonym(s):

L-Lactide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

optical activity

[α]20/D −285°, c = 1 in toluene

mp

92-94 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

C[C@@H]1OC(=O)[C@H](C)OC1=O

InChI

1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3/t3-,4-/m0/s1

InChI key

JJTUDXZGHPGLLC-IMJSIDKUSA-N

Application

(3S)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione may be used in the synthesis of optically active terpyridine containing poly(L-lactide)s, polydisperse lactic acid oligomers, bicyclic and tricyclic lactide derivatives.
Employed in the synthesis of biodegradable copolymers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhang et al.
Polymers, 11(5) (2019-05-15)
To improve the hemocompatibility of the biodegradable medical poly(ether-ester-urethane) (PEEU), containing uniform-size aliphatic hard segments that was prepared in our lab, a copolymer containing phosphorylcholine (PC) groups was blended with the PEEU. The PC-copolymer of poly(MPC-co-EHMA) (PMEH) was first obtained
G Perego et al.
Biomaterials, 15(3), 189-193 (1994-02-01)
A copolymer of L-lactide and 6-caprolactone (50:50, w/w) was synthesized and characterized. The thermal behaviour of this material did not show any crystallinity for several months; only after more than 1 yr of aging at room temperature and, particularly, in
Monodisperse enantiomeric lactic acid oligomers: preparation, characterization, and stereocomplex formation.
De Jong SJ, et al.
Macromolecules, 31(19), 6397-6402 (1998)
Makromol. Chem., 194, 907-907 (1993)
Chaemin Lim et al.
Colloids and surfaces. B, Biointerfaces, 153, 10-18 (2017-02-14)
Polyelectrolyte has been proposed as an efficient approach for various types of drug formulations. However, one drawback of using the conventional polyelectrolyte for drug delivery is its dissociation in in vivo conditions by counter ions due to the lack of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service