Skip to Content
Merck
All Photos(2)

Documents

303143

Sigma-Aldrich

3,6-Dimethyl-1,4-dioxane-2,5-dione

99%

Synonym(s):

DL-Lactide, rac-Lactide, Lactide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

142 °C/8 mmHg (lit.)

mp

116-119 °C

storage temp.

2-8°C

SMILES string

CC1OC(=O)C(C)OC1=O

InChI

1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3

InChI key

JJTUDXZGHPGLLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,6-Dimethyl-1,4-dioxane-2,5-dione (or rac-lactide), is the 50:50 racemic mixture of D- and L-Lactide. Rac-lactide is a lactone derived from lactic acid that has attracted great interest in academia and commercial applications, as it is derived from abundant renewable resources. Rac-lactide can be ready polymerized via ring-opening polymerization, using a variety of metal or organocatalysts, yielding poly(D,L-lactide). While the resulting polymer is generally amorphous, the use of stereospecific catalysts can lead to heterotactic PLA, which exhibits some degree of crystallinity.

Application

3,6-Dimethyl-1,4-dioxane-2,5-dione can be used as a reactant:
  • To synthesize multi-block copolymers of polylactide and polycarbonate.
  • In the aluminum-catalyzed polymerization of propene oxide, lactide, and phthalic anhydride to produce multi-block polyesters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 6517-6517 (2006)
Vibrational Optical Activity of (3 S, 6 S)-3, 6-Dimethyl-1, 4-dioxane-2, 5-dione.
Tam CN, et al.
Journal of the American Chemical Society, 118(42), 10285-10293 (1996)
Lu Qin et al.
Dalton transactions (Cambridge, England : 2003), 48(32), 12315-12325 (2019-07-26)
In this paper we report a series of Al(iii) complexes supported by N,O-bidentate β-pyrazyl functionalized enolate ligands HL1-HL5 (L = (6-Me-2,5-C4H2N2)-CH[double bond, length as m-dash]C(R)-O-), (R = tBu, Ph, p-tolyl, p-OMePh, o-tolyl) and their exploitation for the ring-opening polymerization of
Hendrik Vögeling et al.
Materials science & engineering. C, Materials for biological applications, 103, 109749-109749 (2019-07-28)
Catheter related infections are causing one third of all blood stream infections. The mortality of those infections is very high and the gold standard for catheter related blood stream infections (CR-BSI) is still the removal of the catheter and systemic
Insun Yu et al.
Journal of the American Chemical Society, 134(30), 12758-12773 (2012-07-07)
A family of racemic and enantiopure indium complexes 1-11 bearing bulky chiral diaminoaryloxy ligands, H(NNO(R)), were synthesized and fully characterized. Investigation of both the mono- and the bis-alkoxy-bridged complexes [(NNO(R))InX](2)[μ-Y][μ-OEt] (5, R = (t)Bu, X = Y = Cl; 8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service