Skip to Content
Merck
All Photos(1)

Key Documents

366978

Sigma-Aldrich

5-Fluoroisatin

98%

Synonym(s):

5-Fluoro-2,3-indoledione, NSC 39161

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4FNO2
CAS Number:
Molecular Weight:
165.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

224-227 °C (lit.)

functional group

fluoro
ketone

SMILES string

Fc1ccc2NC(=O)C(=O)c2c1

InChI

1S/C8H4FNO2/c9-4-1-2-6-5(3-4)7(11)8(12)10-6/h1-3H,(H,10,11,12)

InChI key

GKODDAXOSGGARJ-UHFFFAOYSA-N

General description

5-Fluoroisatin has been reported as the precursor of the Sunitinib (Sutent) drug. 5-Fluoroisatin has been approved by the Food and Drugs Administration (FDA) in 2006 for the treatment of renal cell carcinoma (RCC) and gastrointestinal stromal tumor (GIST).

Application

5-Fluoroisatin may be used:
  • as reaction-based probe for live-cell detection of peroxynitrite by 19F magnetic resonance spectroscopy
  • in non-invasive detection of peroxynitrite (ONOO(-)) formation in living lung epithelial cells stimulated with interferon-γ (IFN-γ)
  • in the synthesis of bis-Schiff bases, via condensation with aromatic primary bis-amines in water suspension medium without using any organic solvent or acid catalyst
  • in the synthesis of 3-acetonyl-5-fluoro-3-hydroxyoxindole
Reactant for preparation of:
  • Spiro[indole-thiazolidinones] as biologically relevan synthesis scaffolds
  • Potential antimycobacterial agents
  • Inhibitors of c-Met kinase
  • Inhibitors of TAK1 kinase
  • Herpes simplex virus inhibitors
  • IKKβ inhibitors
  • Inhibitors of vitiligo disease
  • Potential drug candidates with anti-HIV activity and anti-tubercular activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Counter-Current chromatography separation of isatin derivatives using the sandmeyer methodology.
Almeida MR, et al.
Journal of the Brazilian Chemical Society, 21(4), 764-769 (2010)
Kevin J Bruemmer et al.
Chemical communications (Cambridge, England), 50(82), 12311-12314 (2014-09-03)
We report a newly discovered oxidative decarbonylation reaction of isatins that is selectively mediated by peroxynitrite (ONOO(-)) to provide anthranilic acid derivatives. We have harnessed this rapid and selective transformation to develop two reaction-based probes, 5-fluoroisatin and 6-fluoroisatin, for the
A Novel Preparation of a-Substituted Tryptamines from Isatins.
Franklin CS and White AC.
Journal of the Chemical Society, 2, 1335-1337 (1963)
A A Jarrahpour et al.
Molecules (Basel, Switzerland), 11(1), 59-63 (2007-10-27)
Condensation of aromatic primary bis-amines with isatin (1H-indole-2,3-dione) and 5-flouroisatin occurred cleanly and efficiently in a water suspension medium without using any organic solvent or acid catalyst. The corresponding bis-Schiff bases were obtained in good yields and were easily isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service