O9808
2-Oxindole
97%
Synonym(s):
2-Indolinone, Oxindole
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
Beilstein:
114692
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
crystals
bp
227 °C/73 mmHg (lit.)
mp
123-128 °C (lit.)
SMILES string
O=C1Cc2ccccc2N1
InChI
1S/C8H7NO/c10-8-5-6-3-1-2-4-7(6)9-8/h1-4H,5H2,(H,9,10)
InChI key
JYGFTBXVXVMTGB-UHFFFAOYSA-N
Gene Information
human ... PGR(5241)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R Carpenedo et al.
Journal of neurochemistry, 70(5), 1998-2003 (1998-05-08)
Rats treated with oxindole (10-100 mg/kg i.p.), a putative tryptophan metabolite, showed decreased spontaneous locomotor activity, loss of the righting reflex, hypotension, and reversible coma. Brain oxindole levels were 0.05 +/- 0.01 nmol/g in controls and increased to 8.1 +/-
Oxindole in pathogenesis of hepatic encephalopathy.
F Moroni et al.
Lancet (London, England), 351(9119), 1861-1861 (1998-07-04)
Tao Zhang et al.
Chemical communications (Cambridge, England), 49(16), 1636-1638 (2013-01-24)
An enantioselective direct α-alkylation of 2-oxindoles with Michler's hydrol via an S(N)1-type pathway in the non-covalent activation mode using the bis-cinchona alkaloid and Brønsted acid as a co-catalyst was developed and good to high yields and enantioselectivities were obtained.
P B Bergqvist et al.
Pharmacology & toxicology, 85(3), 138-143 (1999-10-16)
It has previously been shown that the neurodepressant L-tryptophan metabolite oxindole is increased in the blood and brain of rats with fulminant hepatic failure and in the blood of cirrhotic patients affected by chronic hepatic encephalopathy. In the present investigation
Zhigang Yang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(22), 6632-6637 (2010-04-22)
A highly enantioselective alpha-amination of 3-substituted oxindoles with azodicarboxylates catalyzed by a chiral Sc(OTf)(3)/N,N'-dioxide complex (Tf: triflate) has been developed and affords the corresponding 3-amino-2-oxindole derivatives in high yields (up to 98%) with excellent enantioselectivities (up to 99% ee). The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service