Skip to Content
Merck
All Photos(1)

Documents

364940

Sigma-Aldrich

Benzene-D6

99.6 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.6 atom % D

Assay

99% (CP)

form

liquid

contains

0.03 % (v/v) TMS

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene-d6 (C6D6) is deuterated benzene. Its Soret coefficient S(T) has been measured by transient holographic grating technique. Its synthesis,13C NMR, IR and MS spectra have been reported.

Application

Benzene-d6 (C6D6) may be used in the following:
  • Synthesis of isotopically labeled samples.
  • As a 1H and 13C-NMR solvent.
  • In the internal calibration of benzene stock solution.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomonitoring of benzene and toluene in human blood by headspace-solid-phase microextraction.
Schimming E, et al.
Fresenius Journal of Analytical Chemistry, 363(1), 88-91 (1999)
Structure-property relationships in nematic gold nanoparticles.
Cseh L and Mehl GH.
Journal of Materials Chemistry, 17(4), 311-315 (2007)
Surprising Titanium complexes bearing η2-pyrazolato ligands: synthesis, structure, and molecular orbital studies.
Guzei IA, et al.
Journal of the American Chemical Society, 119(14), 3387-3388 (1997)
Synthesis and Spectral Analysis of Benzene-d6.
Guo YZ, et al.
Advanced Materials Research, 1061-1062, 301-306 (2014)
The conformational distribution in diphenylmethane determined by nuclear magnetic resonance spectroscopy of a sample dissolved in a nematic liquid crystalline solvent.
Celebre G, et al.
J. Chem. Phys. , 118(14), 6417-6426 (2003)

Related Content

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service