Skip to Content
Merck
All Photos(1)

Documents

175978

Sigma-Aldrich

Benzene-D6

99 atom % D

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Assay

≥99.00%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene-d6 (C6D6) is a deuterated derivative of benzene. Its Soret coefficient S(T) has been measured by the transient holographic grating technique. Its first resonance Raman spectrum and infrared absorption intensities (in the range of 5000-450cm-1) have been reported. It participates in the synthesis of isotopically labeled samples.

Application


  • 2H NMR study of dynamics of benzene-d6 interacting with humic and fulvic acids.: This study examines the interactions of benzene-d6 with humic and fulvic acids using deuterium NMR, highlighting the compound′s utility in probing the structure and dynamics of natural organic matter in environmental contexts (Nanny et al., 2011).


  • Separation of benzene and deuterated benzenes by reversed-phase and recycle liquid chromatography using monolithic capillary columns.: This research demonstrates the effective separation of benzene-d6 from its isotopologues using advanced chromatographic techniques, contributing to methodologies in analytical chemistry for precise isotopic analysis (Lim et al., 2004).


  • Structure-function relationships in high-density docosylsilane bonded stationary phases by Raman spectroscopy and comparison to octadecylsilane bonded stationary phases: effects of aromatic compounds.: The interaction of benzene-d6 with various stationary phases is analyzed, providing insights into the molecular interactions that influence chromatographic separations (Liao & Pemberton, 2008).


  • Reaction of molecular oxygen with a Pd(II)-hydride to produce a Pd(II)-hydroperoxide: experimental evidence for an HX-reductive-elimination pathway.: This study uses benzene-d6 in experimental setups to elucidate mechanisms of oxygen interaction with Pd(II)-hydride complexes, significant in catalytic oxidation processes (Konnick & Stahl, 2008).


  • Quantitative analysis of benzene by selected ion monitoring/gas chromatography/mass spectrometry.: Benzene-d6 is used as an internal standard to develop a highly precise method for quantifying benzene, crucial for environmental monitoring and industrial hygiene (Gruenke et al., 1986).


Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Mi Kim et al.
Food chemistry, 293, 278-284 (2019-06-04)
A headspace sampling-gas chromatography/mass spectrometry (HS-GC/MS) method using mild HS conditions (40 °C, 30 min) was established, validated in terms of specificity, linearity (1.75-87.65 ng mL-1), precision (0.3-9.1% RSD), and accuracy (81.1-117.7%); and applied for the monitoring of 900 commercial beverage samples of six
Resonance Raman scattering of benzene and benzene-d6 with 212.8 nm excitation.
Ziegler LD and Hudson B.
J. Chem. Phys. , 74(2), 982-992 (1981)
The conformational distribution in diphenylmethane determined by nuclear magnetic resonance spectroscopy of a sample dissolved in a nematic liquid crystalline solvent.
Celebre G, et al.
J. Chem. Phys. , 118(14), 6417-6426 (2003)
Infrared intensities of liquids XXII: Optical and dielectric constants, molar polarizabilities, and integrated intensities of liquid benzene-d6 at 25?C between 5000 and 450 cm-1.
Bertie JE and Keefe CD.
Fresenius Journal of Analytical Chemistry, 362(1), 91-108 (1998)
G Wittko et al.
The Journal of chemical physics, 123(1), 014506-014506 (2005-07-23)
The Soret coefficients S(T) of the liquids acetone, benzene, benzene-d1, 1,3,5-benzene-d3, benzene-d5, benzene-13C6, benzene-d6, n-hexane, toluene, 1,2,3,4-tetrahydronaphthalene, isobutylbenzene, and 1,6-dibromohexane have been measured in protonated and perdeuterated cyclohexane by a transient holographic grating technique. It has been found that S(T)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service