Skip to Content
Merck
All Photos(2)

Key Documents

348325

Sigma-Aldrich

Boron trichloride solution

1.0 M in heptane

Synonym(s):

Boron chloride, Trichloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

1.0 M in heptane

density

0.74 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClB(Cl)Cl

InChI

1S/BCl3/c2-1(3)4

InChI key

FAQYAMRNWDIXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Boron trichloride, a Lewis acid, is a general reagent for the cleavage of a wide range of ether and acetal protecting groups.
  • It is a useful reagent to directly convert aromatic aldehydes to the corresponding gem-dichlorides.
  • It can also be used in the transmetallation reaction with less nucleophilic reagents such as tin and zirconium organometallic compounds to synthesize a variety of organoboranes.

For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Migration of 1-alkenyl groups from zirconium to boron compounds.
Cole T E, et al.
Organometallics, 10(10), 3777-3781 (1991)
Boron Trichloride
Miyaura N, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2006)
Impact of Different Environmental Stimuli on the Release of 1-MCP from Boron-MCP Complexes.
Shahrin T, et al.
Journal of Plant Studies, 6(1), 46-46 (2016)
Conversion of aromatic aldehydes to gem-dichlorides using boron trichloride. A new highly efficient method for preparing dichloroarylmethanes.
Kabalka G W and Wu Z
Tetrahedron Letters, 41(5), 579-581 (2000)
Jayaraman Selvakumar et al.
Organic & biomolecular chemistry, 8(18), 4056-4058 (2010-07-29)
Isoindoloisoquinalinone, pyrroloisoquinolinone and benzo[a]quinolizinone units are constructed via intramolecular cyclization of the methoxy substituted N-phenethylimides using BBr(3).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service