Skip to Content
Merck
All Photos(2)

Key Documents

343625

Sigma-Aldrich

7-Oxooctanoic acid

98%

Synonym(s):

7-Ketooctanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO(CH2)5CO2H
CAS Number:
Molecular Weight:
158.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

160-162 °C/4 mmHg (lit.)

mp

27-29 °C (lit.)

functional group

carboxylic acid
ketone

SMILES string

CC(=O)CCCCCC(O)=O

InChI

1S/C8H14O3/c1-7(9)5-3-2-4-6-8(10)11/h2-6H2,1H3,(H,10,11)

InChI key

OSAHCBHKCKPJGI-UHFFFAOYSA-N

General description

7-Oxooctanoic acid is fatty acid formed by the action of acid synthases from acetyl-CoA and malonyl-CoA precursors.

Application

7-Oxooctanoic acid was employed as test compound to investigate the applicability of high-resolution electrospray ionization mass spectrometry to the measurements of average oxygen to carbon ratio (O/C) in secondary organic aerosols. It may be used in the synthesis of hapten, which was used to obtain antibody for the detection of triacetone triperoxide (an improvised explosive). It may be used in the synthesis of “queen substance” of the honey bee.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Astrid Walter et al.
Langmuir : the ACS journal of surfaces and colloids, 26(19), 15418-15423 (2010-09-08)
Triacetone triperoxide (TATP), an improvised explosive, is a potential security threat because of its cost-efficient synthesis and the difficulty in detecting it. A highly selective antibody could provide the necessary specificity to the detection process. To obtain antibodies, a hapten
Adam P Bateman et al.
Environmental science & technology, 46(15), 8315-8324 (2012-07-04)
The applicability of high-resolution electrospray ionization mass spectrometry (HR ESI-MS) to measurements of the average oxygen to carbon ratio (O/C) in secondary organic aerosols (SOAs) was investigated. Solutions with known average O/C containing up to 10 standard compounds representative of
O A Mamer et al.
Biomedical mass spectrometry, 7(2), 53-57 (1980-02-01)
A patient with recurrent severe hypoglycemia resembling Reye's syndrome was found to have large accumulations of omega -- 1 hydroxy and keto acids in serum and urine that persisted following clinical recovery. A deficiency of mitochondrial medium chain acyl CoA
A Two-Step Synthesis of the ?Queen Substance? of the Honey Bee.
Ebert GW.
Synthetic Communications, 21(14), 1527-1531 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service