Skip to Content
Merck
All Photos(1)

Key Documents

342289

Sigma-Aldrich

cis,cis-1,3,5-Trimethylcyclohexane-1,3,5-tricarboxylic acid

99%

Synonym(s):

Kemp’s triacid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O6
CAS Number:
Molecular Weight:
258.27
Beilstein:
3557263
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

241-243 °C (lit.)

SMILES string

C[C@@]1(C[C@](C)(C[C@](C)(C1)C(O)=O)C(O)=O)C(O)=O

InChI

1S/C12H18O6/c1-10(7(13)14)4-11(2,8(15)16)6-12(3,5-10)9(17)18/h4-6H2,1-3H3,(H,13,14)(H,15,16)(H,17,18)/t10-,11+,12-

InChI key

AZWHPGZNOIYGFB-ZSBIGDGJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile molecule for molecular recognition studies. Also used to prepare a chiral auxiliary for asymmetric radical addition, allylation, and annulation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 1919-1919 (1992)
Journal of the American Chemical Society, 114, 7707-7707 (1992)
J. Chem. Soc. Perkin Trans. II, 569-569 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service