Skip to Content
Merck
All Photos(1)

Key Documents

430595

Sigma-Aldrich

Ytterbium(III) trifluoromethanesulfonate

99.99%

Synonym(s):

Yb(OTf)3, Trifluoromethanesulfonic acid ytterbium(III) salt, Yb(TFA)3, Ytterbium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Yb
CAS Number:
Molecular Weight:
620.25
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.99%

reaction suitability

core: ytterbium
reagent type: catalyst

SMILES string

FC(F)(F)S(=O)(=O)O[Yb](OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Yb/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

AHZJKOKFZJYCLG-UHFFFAOYSA-K

Application

A water-tolerant Lewis acid catalyst recently used in one-pot syntheses of β-lactams, and in stereocontrolled radical and nucleophilic addition reactions. Catalyzes the synthesis of deoxypenostatin A in a novel, stereoselective, intramolecular Diels-Alder reaction.
Lewis acid catalyst used with a silylated chinchona alkaloid in an asymmetric synthesis of substituted β-lactams from an acid chloride and aryl imine. Also used to catalyze an aldehyde-ene reaction in a synthesis of the guaiane skeleton.
An important Lewis acid catalyst for a variety of synthetic reactions, including allylborations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kobayashi, S.
Transition Metals for Organic Synthesis, 1, 285-285 (1998)
Acyclic Stereocontrol in Radical Reactions. Diastereoselective Radical Addition/Allylation of N-Propenoyloxazolidinone.
Mukund P. Sibi et al.
The Journal of organic chemistry, 61(18), 6090-6091 (1996-09-06)
Jili Li et al.
Organic & biomolecular chemistry, 16(43), 8115-8129 (2018-10-20)
CO-releasing molecules (CORMs) containing cobalt have many bioactivities, but most of them do not dissolve in water and have no selectivity to tissue and organs. On the basis of the specific recognition of galactose or sialic acid by a receptor
Jason W J Kennedy et al.
The Journal of organic chemistry, 69(13), 4412-4428 (2004-06-19)
A full account of the development of the first catalytic manifold for the additions of allylboronates to aldehydes is described. The thermal additions (both diastereospecific and enantioselective) of 2-carboxyester 3,3-disubstituted allylboronates 1 to both aromatic and aliphatic aldehydes give biologically
Synlett, 3411-3411 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service