Skip to Content
Merck
All Photos(3)

Key Documents

339202

Sigma-Aldrich

Niobium(V) ethoxide

99.95% trace metals basis

Synonym(s):

NbOEt, Niobium ethanolate, Niobium ethylate, Niobium pentaethoxide, Pentaethoxyniobium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nb(OCH2CH3)5
CAS Number:
Molecular Weight:
318.21
Beilstein:
3678883
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.95% trace metals basis

form

liquid

reaction suitability

core: niobium

refractive index

n20/D 1.516 (lit.)

bp

142 °C/0.1 mmHg (lit.)

mp

5-6 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

SMILES string

CCO[Nb](OCC)(OCC)(OCC)OCC

InChI

1S/5C2H5O.Nb/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5

InChI key

ZTILUDNICMILKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Niobium(V) ethoxide is a metal-organic compound that dimerizes in solution to form Nb2(OEt)10 molecules. At room temperature, it is a colorless liquid that is miscible with some organic solvents, but which hydrolyzes readily. Niobium(V) ethoxide freezes at 5 °C and boils at 142 °C at 0.1 mmHg. It can be synthesized by the electrochemical reaction of ethanol with niobium plate and stainless-steel electrodes in the presence of tetraethylammonium chloride (TEAC) as the conductive additive.

Application

Niobium(V) ethoxide is a synthetic precursor for niobium oxides, for example Nb2O5 , NaNbO3 , andTiNb2O7. Niobium(V) ethoxide is commonly used in colloidal solution-based syntheses like sol-gel processing and electrospinning. These chemistries allow researchers to control the microstructure to form gels , nanofibers , nanotubes , and microspheres. Niobium(V) ethoxide is also commonly added to other metal alkoxides to dope metal oxides, most commonly titania, with niobium.

Because of its low boiling point and low decomposition temperature (~325-350 °C),niobium(V) ethoxide is also used as a precursor in atomic-layer deposition, chemical vapor deposition, and metal-organic chemical vapor deposition of niobium oxides.

It can also be used as a dopant to prepare Nb-doped TiO2 photoanode for dye-sensitized solar cells (DSSC). The doping of Nb lowers the conduction band of TiO2 enhancing electron injection and transfer ability of TiO2 photoanode. This results in a significant increase in the performance of DSSC.

Features and Benefits

The purity and volatility of Niobium(V) ethoxide makeit a suitable precursor for atomic layer deposition and chemical vapordeposition of niobium oxide.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Materials Research Society Monographs, 5, 93-100 (1992)
Synthesis of hierarchical porous TiNb2O7 nanotubes with controllable porosity and their application in high power Li-ion batteries
Park H, et al.
Journal of Material Chemistry A, 5, 6958-6965 (2017)
Porosity-Controlled TiNb2O7 Microspheres with Partial Nitridation as A Practical Negative Electrode for High-Power Lithium-Ion Batteries.
Park H, et al.
Advanced Energy Materials, 5, 1401945-1401945 (2015)
Sol?gel synthesis and characterization of Nb2O5 powders.
Ristic M, et al.
Materials Letters, 58, 2658-2663 (2004)
TiNb2O7 hollow nanofiber anode with superior electrochemical performance in rechargeable lithium ion batteries.
Yu H, et al.
Nano Energy, 38, 109-117 (2017)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service