Skip to Content
Merck
All Photos(1)

Key Documents

87560

Sigma-Aldrich

Titanium(IV) isopropoxide

≥97.0%

Synonym(s):

TTIP, Tetraisopropyl orthotitanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti[OCH(CH3)2]4
CAS Number:
Molecular Weight:
284.22
Beilstein:
3679474
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0%

form

liquid

reaction suitability

core: titanium
reagent type: Lewis acid
reagent type: catalyst

refractive index

n20/D 1.464 (lit.)

bp

232 °C (lit.)

mp

14-17 °C (lit.)

density

0.96 g/mL at 20 °C (lit.)

SMILES string

CC(C)O[Ti](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/4C3H7O.Ti/c4*1-3(2)4;/h4*3H,1-2H3;/q4*-1;+4

InChI key

VXUYXOFXAQZZMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) isopropoxide is a Lewis acid catalyst used in organic reactions like Sharpless epoxidation.[1]

Application

Novel metal oxide/phosphonate hybrids were formed from titanium(IV) isopropoxide in a two-step sol-gel process. Starting material for barium-strontium-titanate thin films. Used to make porous titanosilicates, potential ion-exchange materials for cleanup of radioactive wastes. Applied in the formation of a heterosupermolecule consisting of a TiO2 nanocrystallite-viologen electron acceptor complex whose light-induced electron transfer has been demonstrated.

Caution

may discolor on storage

Other Notes

Additive used in the Sharpless asymmetric epoxidation of allylic alcohols[2]; Catalyst used for transesterifications with various alcohols under neutral conditions[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F - Pensky-Martens closed cup

Flash Point(C)

41 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Imwinkelried et al.
Organic Syntheses, 65, 230-230 (1987)
Catalyst design for alkene epoxidation by molecular analogues of heterogeneous titanium-silicalite catalysts
Sole DA, et al.
ACS Catalysis, 10, 4737-4750 (2020)
Tadashi Kokubo et al.
Journal of materials science. Materials in medicine, 19(2), 695-702 (2007-07-11)
A rectangular specimen of polyethylene terephthalate (PET) was soaked in a titania solution composed of titanium isopropoxide, water, ethanol and nitric acid at 25 degrees C for 1 h. An amorphous titanium oxide was formed uniformly on the surface of
Yann Sarazin et al.
Dalton transactions (Cambridge, England : 2003), (2)(2), 340-350 (2005-12-21)
The reactions of the bulky amino-bis(phenol) ligand Me(2)NCH(2)CH(2)N[CH(2)-3,5-Bu(t)(2)-C(6)H(2)OH-2](2)(1-H(2)) with Zn[N(SiMe(3))(2)](2)(4), [Mg[N(SiMe(3))(2)](2)](2)(5) and Ca[N(SiMe(3))(2)](2)(THF)(2)(6) yield the complexes 1-Zn, 1-Mg and 1-Ca in good yields. The X-ray structure of 1-Ca showed the complex to be dimeric, with calcium in a distorted octahedral
Henrique Antonio Mendonça Faria et al.
Materials science & engineering. C, Materials for biological applications, 56, 260-268 (2015-08-08)
The structural and electronic properties of titanium oxide nanotubes (TiO2) have attracted considerable attention for the development of therapeutic devices and imaging probes for nanomedicine. However, the fluorescence response of TiO2 has typically been within ultraviolet spectrum. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service