Skip to Content
Merck
All Photos(1)

Key Documents

337455

Sigma-Aldrich

(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

97%

Synonym(s):

[Ir(PMePh2)2COD]+ PF6-, [Ir(cod)(PPh2Me)2]PF6, {Ir(COD)[PCH3(C6H5)2]2}PF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H38F6IrP3
CAS Number:
Molecular Weight:
845.79
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

core: iridium
reagent type: catalyst

mp

224 °C (dec.) (lit.)

SMILES string

[Ir+].F[P-](F)(F)(F)(F)F.C1CC=CCCC=C1.CP(c2ccccc2)c3ccccc3.CP(c4ccccc4)c5ccccc5

InChI

1S/2C13H13P.C8H12.F6P.Ir/c2*1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13;1-2-4-6-8-7-5-3-1;1-7(2,3,4,5)6;/h2*2-11H,1H3;1-2,7-8H,3-6H2;;/q;;;-1;+1/b;;2-1-,8-7-;;

InChI key

LXKHQEQLBSJJCO-JXNOXZOESA-N

Application

(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an air-stable cationic iridium complex, generally employed in allyl isomerization reactions. Upon bubbling hydrogen into the solution of [Ir(cod)(PPh2Me)2]PF6, preferably in tetrahydrofuran, results in the formation of active catalyst [IrH2(solv)2(PPh2Me)2]PF6 which can be used in the isomerization of 3-(silyloxy)-1-propenylboronates to (γ-(silyloxy)allyl)boronic esters; diallyl ethers to allyl (E)-vinyl ethers and 1-alkenylboronates to corresponding 3-alkoxyallylboronates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Intramolecular allylboration of γ-(?-formylalkoxy) allylboronates for syntheses of trans-or cis-2-(ethenyl) tetrahydropyran-3-ol and 2-(ethenyl) oxepan-3-ol.
Yamamoto Y, et al.
Tetrahedron, 59(4), 537-542 (2003)
Stereoselective Isomerization of Unsymmetrical Diallyl Ethers to Allyl (E)-Vinyl Ethers by a Cationic Iridium Catalyst.
Yamamoto Y, et al.
Synthetic Communications, 30(13), 2383-2391 (2000)
Synthesis of Chiral Esters of (E)-3-(Silyloxy)-2-propenylboronic Acid via the Iridium-Catalyzed Isomerization of the Double Bond.
Yamamoto Y, et al.
The Journal of Organic Chemistry, 64(1), 296-298 (1999)
Paulsen, H.; Adermann, K.
Carbohydrate Research, 175, 163-163 (1988)
Isomerisation of allyl ethers catalysed by the cationic iridium complex [Ir (cyclo-octa-1, 5-diene)(PMePh 2) 2] PF 6. A highly stereoselective route to trans-propenyl ethers.
Baudry D, et al.
Journal of the Chemical Society. Chemical Communications, (16), 694-695 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service