Skip to Content
Merck
All Photos(3)

Documents

335991

Sigma-Aldrich

Tetrabutylammonium acetate

97%

Synonym(s):

N,N,N-tributyl-1-butanaminium acetate, Tetra-n-butylammonium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCOCH3)
CAS Number:
Molecular Weight:
301.51
Beilstein:
3599376
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

95-98 °C (lit.)

SMILES string

CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C2H4O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2(3)4/h5-16H2,1-4H3;1H3,(H,3,4)/q+1;/p-1

InChI key

MCZDHTKJGDCTAE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium acetate (TBAA) is a phase transfer catalyst (PTC).

Application

Tetrabutylammonium acetate can be used:
  • To catalyze the alkynylation of carbonyl compounds with trimethylsilylacetylenes providing good yields of propargylic alcohols.
  • Along with tetrabutylammonium bromide (TBAB) as a molten reaction medium in the synthesis of 4-aryl-2-quinolones.
  • As a promoter in the regioselective direct arylation of azoles (1-methylpyrazole, oxazole, and thiazole) with aryl bromides in presence of palladium acetate catalyst.
  • As an ionic liquid to dissolve cellulose in the presence of DMSO co-solvent.
  • As an activator in the synthesis of disubstituted alkynes through copper-free Sonogashira coupling reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4?Aryl?2?quinolones through a Pseudo?Domino Heck/Buchwald?Hartwig Reaction in a Molten Tetrabutylammonium Acetate/Tetrabutylammonium Bromide Mixture
Battistuzzi G, et al.
advanced synthesis and catalysis, 3(349), 297-302 (2007)
Palladium-based catalytic systems for the synthesis of conjugated enynes by Sonogashira reactions and related alkynylations
Doucet H, and Hierso J-C,
Angewandte Chemie (International Edition in English), 46(6), 834-871 (2007)
Quaternary ammonium acetate: an efficient ionic liquid for the dissolution and regeneration of cellulose
Miao J, et al.
Royal Society of Chemistry Advances, 4(69), 36721-36724 (2014)
Chemistry Letters (Jpn), 35, 1002-1002 (2006)
Mild Palladium?Catalyzed Regioselective Direct Arylation of Azoles Promoted by Tetrabutylammonium Acetate
Bellina F, et al.
European Journal of Organic Chemistry, 25(349), 5621-5630 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service