Skip to Content
Merck
All Photos(2)

Key Documents

245070

Sigma-Aldrich

Tetramethylammonium acetate

≥97%

Synonym(s):

N,N,N-trimethyl-methanaminium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OCOCH3)
CAS Number:
Molecular Weight:
133.19
Beilstein:
3571830
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

solid

mp

184 °C (dec.) (lit.)

SMILES string

CC([O-])=O.C[N+](C)(C)C

InChI

1S/C4H12N.C2H4O2/c1-5(2,3)4;1-2(3)4/h1-4H3;1H3,(H,3,4)/q+1;/p-1

InChI key

MRYQZMHVZZSQRT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra M Urbanska et al.
Cancer biotherapy & radiopharmaceuticals, 35(6), 459-473 (2020-02-06)
An α particle-emitting nanodrug that is a potent and specific antitumor agent and also prompts significant remodeling of local immunity in the tumor microenvironment (TME) has been developed and may impact the treatment of melanoma. Biocompatible ultrasmall fluorescent core-shell silica
Mesude Bicak et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(26), 15172-15181 (2020-06-14)
Hu11B6 is a monoclonal antibody that internalizes in cells expressing androgen receptor (AR)-regulated prostate-specific enzyme human kallikrein-related peptidase 2 (hK2; KLK2). In multiple rodent models, Actinium-225-labeled hu11B6-IgG1 ([225Ac]hu11B6-IgG1) has shown promising treatment efficacy. In the present study, we investigated options
Zhiyuan Zhao et al.
ACS omega, 5(30), 19247-19254 (2020-08-11)
In this work, poly(ethylene terephthalate) (PET) chain-extending products with different molecular weights were prepared by reactive extrusion using isocyanate trimer (C-HK) as the trifunctional chain extender. The effect of the chain extender C-HK on the intrinsic viscosity, melt flow property
Daniel L J Thorek et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(2), 881-891 (2018-09-27)
The impact of androgen receptor (AR) activity in breast cancer biology is unclear. We characterized and tested a novel therapy to an AR-governed target in breast cancer.Experimental Design: We evaluated the expression of prototypical AR gene products human kallikrein 2
Alexey A Nazarov et al.
Dalton transactions (Cambridge, England : 2003), 44(8), 3614-3623 (2014-11-20)
Chemotherapeutics for the treatment of tumorigenic conditions that feature novel modes of action are highly sought after to overcome the limitations of current chemotherapies. Herein, we report the conjugation of the alkylating agent chlorambucil to the RAPTA scaffold, a well-established

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service