Skip to Content
Merck
All Photos(3)

Key Documents

335355

Sigma-Aldrich

Coronene

purified by sublimation, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H12
CAS Number:
Molecular Weight:
300.35
Beilstein:
658468
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

purified by

sublimation

bp

525 °C (lit.)

mp

428 °C (lit.)

SMILES string

c1cc2ccc3ccc4ccc5ccc6ccc1c7c2c3c4c5c67

InChI

1S/C24H12/c1-2-14-5-6-16-9-11-18-12-10-17-8-7-15-4-3-13(1)19-20(14)22(16)24(18)23(17)21(15)19/h1-12H

InChI key

VPUGDVKSAQVFFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An n-channel organic semiconductor.
Used to synthesize MBE-grown layered superconductors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jpn J. App. Phys., Part I, 34, 3837-3845 (1995)
Hirofumi Sato et al.
Physical chemistry chemical physics : PCCP, 13(1), 309-313 (2010-11-26)
Coronene (C(24)H(12)) is a flat polyaromatic hydrocarbon consisting of seven peri-fused benzene rings and attracts lots of attention as a fragment of graphene. Using a hybrid method of quantum chemistry and statistical mechanics called RISM-SCF, which is an alternative to
Anna M Hiszpanski et al.
ACS nano, 7(1), 294-300 (2012-12-12)
The structuring in organic electrically active thin films critically influences the performance of devices comprising them. Controlling film structure, however, remains challenging and generally requires stringent deposition conditions or modification of the substrate. To this end, we have developed post-deposition
Seok Ju Kang et al.
Angewandte Chemie (International ed. in English), 51(34), 8594-8597 (2012-07-19)
"Ball and socket" motif: The contorted dibenzotetrathienocoronene (6-DBTTC) forms a complex with the C(70) fullerene PC(70) BM embedded in an amorphous phase of PC(70) BM. The materials are processable into organic solar cells in solution. The power conversion efficiency is
Long Chen et al.
Journal of the American Chemical Society, 134(43), 17869-17872 (2012-10-16)
Here we report hexathienocoronenes (HTCs), fully thiophene-annelated coronenes in which six double bonds in the periphery are thieno-fused. The derivatives tetrasubstituted with hexyl and dodecyl chains show a phase formation that strongly depends on the chain length. HTCs are remarkably

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service