Skip to Content
Merck
All Photos(1)

Key Documents

328103

Sigma-Aldrich

3′-Hydroxyacetophenone

≥99%

Synonym(s):

1-(3-Hydroxyphenyl)ethanone, 3-Acetylphenol, 3-Hydroxyphenylethanone, m-Hydroxyacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
2040676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

bp

296 °C (lit.)

mp

90-95 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cccc(O)c1

InChI

1S/C8H8O2/c1-6(9)7-3-2-4-8(10)5-7/h2-5,10H,1H3

InChI key

LUJMEECXHPYQOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3′-Hydroxyacetophenone is an hydroxy-substituted alkyl phenyl ketone.[1]

Application

3′-Hydroxyacetophenone was used in synthesis of enantiopure (-)-rivastigmine.[2] It was also used in preparation of building block for synthesis of dendritic compounds.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

247.3 °F - closed cup

Flash Point(C)

119.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of polyester dendrimers and dendrons starting from Michael reaction of acrylates with 3-hydroxyacetophenone.
Hirayama Y, et a
Tetrahedron Letters, 46(6), 1027-1030 (2005)
Kiwon Han et al.
The Journal of organic chemistry, 75(9), 3105-3108 (2010-03-30)
A practical and efficient procedure for the synthesis of rivastigmine was developed. This procedure includes dynamic kinetic resolution using a polymer-bound ruthenium complex and a lipase in combination as a key step. Enantiopure (-)-rivastigmine was obtained from commercially available 3'-hydroxyacetophenone
Katharine Moore Tibbetts et al.
The journal of physical chemistry. A, 118(37), 8170-8176 (2014-03-01)
The hydroxy-substituted alkyl phenyl ketones 2'-, 3'- and 4'- (ortho, meta, and para) hydroxyacetophenone were excited in the strong-field regime with wavelengths ranging from 1200-1500 nm to produce the respective radical cations. For 2'- and 3'-hydroxyacetophenone, the parent molecular ion
Takashi Otani et al.
Bioorganic & medicinal chemistry letters, 18(12), 3582-3584 (2008-05-31)
m-Acetylphenyl-beta-d-glucopyranosides and m-acetylphenyl-alpha/beta-d-mannopyranosides were synthesized by the Koenigs-Knorr, Mitsunobu, and Helferich reactions as key glycosylation reactions, respectively. Their spectroscopic properties and antioxidative activities were characterized as potential ultraviolet B-ray absorbents.
Stefania Ferrari et al.
Journal of medicinal chemistry, 54(1), 211-221 (2010-12-04)
Folate analogue inhibitors of Leishmania major pteridine reductase (PTR1) are potential antiparasitic drug candidates for combined therapy with dihydrofolate reductase (DHFR) inhibitors. To identify new molecules with specificity for PTR1, we carried out a virtual screening of the Available Chemicals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service