Skip to Content
Merck
All Photos(2)

Documents

325872

Sigma-Aldrich

Samarium(III) acetate hydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Sm · xH2O
CAS Number:
Molecular Weight:
327.49 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

core: samarium
reagent type: catalyst

SMILES string

O.CC(=O)O[Sm](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.H2O.Sm/c3*1-2(3)4;;/h3*1H3,(H,3,4);1H2;/q;;;;+3/p-3

InChI key

LKSVMAKMHXTSQX-UHFFFAOYSA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masao Nakayama et al.
Physica medica : PM : an international journal devoted to the applications of physics to medicine and biology : official journal of the Italian Association of Biomedical Physics (AIFB), 75, 69-76 (2020-06-17)
Titanium dioxide nanoparticles (TiO2 NPs) have been investigated for their role as radiosensitisers for radiation therapy. The study aims to increase the efficiency of these NPs by synthesising them with samarium. Samarium-doped TiO2 NPs (Ti(Sm)O2 NPs) were synthesised using a
Chao Wang et al.
DNA repair, 95, 102946-102946 (2020-08-28)
Ataxia Telangiectasia and Rad3-Related kinase (ATR) is a master regulator of genome maintenance, and participates in DNA replication and various DNA repair pathways. In a genome-wide screen for ATR-dependent fitness genes, we identified a previously uncharacterized gene, C17orf53, whose loss
Xixi Wang et al.
Nanoscale, 12(36), 18807-18814 (2020-09-25)
Mechanistic studies on photon upconversion play a critical role in the fundamental research of the luminescence of rare earth ions as well as their emerging applications. Energy migration mediated upconversion (EMU) has recently shown to be an important model for
Amit K Guria et al.
Scientific reports, 4, 6514-6514 (2014-10-02)
Programming the reaction chemistry for superseding the formation of Sm2O3 in a competitive process of formation and dissolution, the crystal growth patterns are varied and two different nanostructures of Sm2O3 in 2D confinement regime are designed. Among these, the regular
Bo Zhou et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700667-1700667 (2018-03-30)
Upconverting materials have achieved great progress in recent years, however, it remains challenging for the mechanistic research on new upconversion strategy of lanthanides. Here, a novel and efficient strategy to realize photon upconversion from more lanthanides and fine control of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service