Merck
  • Tuning the growth pattern in 2D confinement regime of Sm2O3 and the emerging room temperature unusual superparamagnetism.
Tuning the growth pattern in 2D confinement regime of Sm2O3 and the emerging room temperature unusual superparamagnetism.
Scientific reports (2014-10-02)
Amit K Guria, Koushik Dey, Suresh Sarkar, Biplab K Patra, Saurav Giri, Narayan Pradhan
ABSTRACT
Programming the reaction chemistry for superseding the formation of Sm2O3 in a competitive process of formation and dissolution, the crystal growth patterns are varied and two different nanostructures of Sm2O3 in 2D confinement regime are designed. Among these, the regular and self-assembled square platelets nanostructures exhibit paramagnetic behavior analogous to the bulk Sm2O3. But, the other one, 2D flower like shaped nanostructure, formed by irregular crystal growth, shows superparamagnetism at room temperature which is unusual for bulk paramagnet. It has been noted that the variation in the crystal growth pattern is due to the difference in the binding ability of two organic ligands, oleylamine and oleic acid, used for the synthesis and the magnetic behavior of the nanostructures is related to the defects incorporated during the crystal growth. Herein, we inspect the formation chemistry and plausible origin of contrasting magnetism of these nanostructures of Sm2O3.
Product Description
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Samarium(III) acetate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
Supelco
Oleic acid, analytical standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard