Skip to Content
Merck
All Photos(7)

Key Documents

791725

Sigma-Aldrich

Citric acid

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%

Synonym(s):

2-Hydroxy-propane-1,2,3-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.5%

form

crystals

quality

free-flowing

expl. lim.

8 %, 65 °F

impurities

Substances carbonizable by hot sulfuric acid, passes test

ign. residue

≤0.02%

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
oxalate (C2O42-): passes test (limit about 0.003%)
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤3 ppm
Pb: ≤2 ppm

functional group

carboxylic acid
hydroxyl

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Citric acid also called 2-hydroxy-propane-1,2,3-tricarboxylic acid, is a component of the tricarboxylic acid cycle in all organisms. Industrial production of citric acid use fungal, yeast and citrus fruit as the major source. Aspergillus niger is majorly used for citric acid production by fermentation Manganese inhibits fermentative production of citric acid Oral supplementation of citric acid promotes tricarboxylic acid cycle and oxidative phosphorylation in mitochondria, attenuating physical fatigue.

Application

Citric acidhas been used as a component of citrate buffer and Fe-free sauton′s medium. It has been used in hybridization chain reaction buffer for preequilibration of coverslips for testes sections.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comprehensive roadmap of murine spermatogenesis defined by single-cell RNA-seq
Green CD, et al.
Developmental Cell, 46(5), 651-667 (2018)
Biotechnological production of citric acid
Max B, et al.
Brazilian Journal of Microbiology, 41(4), 862-875 (2010)
Methylene blue exerts a neuroprotective effect against traumatic brain injury by promoting autophagy and inhibiting microglial activation
Zhao M, et al.
Molecular Medicine Reports, 13(1), 13-20 (2016)
Two promoters in the esx-3 gene cluster of Mycobacterium smegmatis respond inversely to different iron concentrations in vitro
Fang Z, et al.
BMC Research Notes, 10(1), 426-426 (2017)
Effects of citric acid and L-carnitine on physical fatigue
Sugino T, et al.
Journal of Clinical Biochemistry and Nutrition, 41(3), 224-230 (2007)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service