Skip to Content
Merck
All Photos(2)

Key Documents

303585

Sigma-Aldrich

2-Amino-5-nitrophenol

Synonym(s):

2-Hydroxy-4-nitroaniline, 3-Hydroxy-4-aminonitrobenzene, 3-Nitro-6-aminophenol, 5-Nitro-2-amino-1-hydroxybenzene, 5-Nitro-2-aminophenol, NSC 7087

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
972974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

198-202 °C (dec.) (lit.)

SMILES string

Nc1ccc(cc1O)[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-5-2-1-4(8(10)11)3-6(5)9/h1-3,9H,7H2

InChI key

DOPJTDJKZNWLRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-5-nitrophenol was used:
  • as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides
  • as semipermanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products
  • in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents
Reactant for:
  • Diazotation and coupling reactions
  • Preparation of biologically and pharmacologically active molecules

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadeem Siddiqui et al.
Archiv der Pharmazie, 343(11-12), 657-663 (2010-11-27)
A series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides (6a-r) was synthesized using 2-amino-5-nitrophenol as a starting material. All the synthesized compounds possessed two hydrogen-bonding domains and their effect on the activity was studied thereof. The anticonvulsant activity was assessed by
2-Amino-5-nitrophenol.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 177-184 (1993-01-01)
F Chen et al.
Cancer letters, 126(1), 67-74 (1998-05-01)
Two hair dye components, carcinogenic 4-nitro-2-aminophenol and 5-nitro-2-aminophenol, induced Cu(II)-dependent DNA cleavage frequently at thymine and guanine residues in DNA fragments obtained from the c-Ha-ras-1 protooncogene. When the p53 tumor suppressor gene was used, 4-nitro-2-aminophenol caused Cu(II)-dependent piperidine-labile sites at
Christina L Burnett et al.
International journal of toxicology, 28(6 Suppl 2), 217S-251S (2010-01-30)
2-Amino-3-nitrophenol, 2-amino-4-nitrophenol, 2-amino-5-nitrophenol, 4-amino-3-nitrophenol, 4-amino-2-nitrophenol, 2-amino-4-nitrophenol sulfate, 3-nitro-p-hydroxyethylaminophenol, and 4-hydroxypropylamino-3-nitrophenol are substituted aromatic compounds used as semipermanent (nonoxidative) hair colorants and as toners in permanent (oxidative) hair dye products. All ingredients in this group except 2-amino-4-nitrophenol sulfate, 2-amino-5-nitrophenol, and 4-amino-2-nitrophenol
Weixia Zhu et al.
Se pu = Chinese journal of chromatography, 30(9), 870-875 (2013-01-05)
An analytical method was developed for the simultaneous determination of 11 aminophenols in direct and oxidized hair dyes by high performance liquid chromatography (HPLC). The samples were extracted with methanol using sodium bisulfite for anti-oxidation. The purification was carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service