Skip to Content
Merck
All Photos(6)

Key Documents

299251

Sigma-Aldrich

Ammonium-15N chloride

≥98 atom % 15N, ≥99% (CP)

Synonym(s):

15N Labeled ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15NH4Cl
CAS Number:
Molecular Weight:
54.48
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % 15N

Assay

≥99% (CP)

form

powder, crystals or chunks

technique(s)

bio NMR: suitable
protein expression: suitable

mp

340 °C (subl.) (lit.)

mass shift

M+1

storage temp.

room temp

SMILES string

[15NH4+].[Cl-]

InChI

1S/ClH.H3N/h1H;1H3/i;1+1

InChI key

NLXLAEXVIDQMFP-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium-15N chloride has been used:
  • as a medium supplement for the incorporation of 15N in sporulation protein, CsfB, for NMR spectroscopy
  • in assaying nitrogen loss in marine sediment
  • for the incorporation of 15N in apolipoprotein A-I for mass spectrum analysis

Biochem/physiol Actions

Ammonium chloride is a component of the nitrogen cycle. Excess of ammonium chloride depletes ATP in astrocytes. Ammonium chloride inhibits the acidification in endosome-lysosome system. It is a hemolysing agent used in the lysis of erythrocytes. Ammonium chloride administration promotes creatinine and urea clearance in rat.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 46, 4296-4296 (1981)
Zhen-Zhen Zheng et al.
Nature communications, 11(1), 3511-3511 (2020-07-16)
In the context of continuously increasing anthropogenic nitrogen inputs, knowledge of how ammonia oxidation (AO) in the ocean responds to warming is crucial to predicting future changes in marine nitrogen biogeochemistry. Here, we show divergent thermal response patterns for marine
Characterization of homodimer interfaces with cross-linking mass spectrometry and isotopically labeled proteins
Lima DB, et al.
Nature Protocols, 13(3), 431-431 (2018)
Brianna J Klein et al.
Structure (London, England : 1993), 28(1), 105-110 (2019-11-13)
The assembly of human histone acetyltransferase MOZ/MORF complexes relies on the scaffolding bromodomain plant homeodomain (PHD) finger 1 (BRPF1) subunit. The PHD-zinc-knuckle-PHD module of BRPF1 (BRPF1PZP) has been shown to associate with the histone H3 tail and DNA; however, the
Anaerobic ammonium oxidation linked to sulfate and ferric iron reduction fuels nitrogen loss in marine sediments
Rios-Del T, et al.
Biodegradation, 29(5), 429-442 (2018)

Articles

Utilizing ISOGRO® Supplementation of M9 Minimal Media to Enhance Recombinant Protein Expression.

Related Content

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Protein structure analysis aids in identifying disease biomarkers and drug targets crucial for therapeutic treatments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service