Skip to Content
Merck
All Photos(2)

Key Documents

251127

Sigma-Aldrich

Bis(tributyltin)

95%

Synonym(s):

Hexabutyldistannane, Hexabutylditin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3SnSn(CH2CH2CH2CH3)3
CAS Number:
Molecular Weight:
580.11
Beilstein:
3605476
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.512 (lit.)

density

1.148 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)[Sn](CCCC)(CCCC)CCCC

InChI

1S/6C4H9.2Sn/c6*1-3-4-2;;/h6*1,3-4H2,2H3;;

InChI key

REDSKZBUUUQMSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(tributyltin) is utilized in tin-carbon bond formation, desulfurization, and deoxygenation reactions.

Application

Reagent used to stannylate aryl halides for subsequent Stille coupling. Also used in the radioiodination of styrlpyridines.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron, 63, 9465-9465 (2007)
Wenchao Qu et al.
Journal of medicinal chemistry, 50(9), 2157-2165 (2007-04-07)
We report a series of radioiodinated styrylpyridines as single photon emission computed tomography probes for imaging Abeta plaques in the brain of patients with Alzheimer's disease (AD). In vitro binding showed that all of the styrylpyridines displayed very good binding
Hexabutyldistannane
Mitchell TN and Nikonov G
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Tetrahedron Letters, 29, 6893-6893 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service