Skip to Content
Merck
All Photos(3)

Key Documents

285943

Sigma-Aldrich

β-D-Glucose pentaacetate

98%

Synonym(s):

beta-D-Glucose pentaacetate, 1,2,3,4,6-Penta-O-acetyl-β-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
Beilstein:
98851
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

optical activity

[α]20/D +4.2°, c = 1 in chloroform

mp

130-132 °C (lit.)

SMILES string

CC(O)O[C@@H]1[C@@H](COC(C)=O)O[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]1OC(C)=O

InChI

1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3/t12-,13-,14+,15-,16-/m1/s1

InChI key

LPTITAGPBXDDGR-IBEHDNSVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monosaccharide esters: new tools in biomedical research.
W J Malaisse
Molecular genetics and metabolism, 65(2), 129-142 (1998-10-27)
W J Malaisse et al.
Archives of biochemistry and biophysics, 381(1), 61-66 (2000-10-06)
Hepatocytes from fed rats were incubated for 120 min in the presence of alpha-D-[1,2-13C]glucose pentaacetate (1.7 mM), both D-[1,2-13C]glucose (1.7 mM) and acetate (8.5 mM), alpha-D-glucose penta[2-13C]acetate (1.7 mM), or D-[1,2-13C]glucose (8.3 mM). The amounts of 13C-enriched L-lactate and D-glucose
G Ayvaz et al.
Research communications in molecular pathology and pharmacology, 103(1), 83-90 (1999-08-10)
The pentaacetate ester of alpha-D-glucose was recently introduced as a new insulin secretagogue. Its insulinotropic action appears mainly attributable to the catabolism of its hexose moiety in islet cells, but a direct effect of the ester itself upon a receptor
F Ponticelli et al.
Carbohydrate research, 330(4), 459-468 (2001-03-28)
We describe the synthesis of some 3-tert-butyl-4-hydroxyphenyl D-glycopyranosides by reaction of tert-butylhydroquinone with beta-D-pentaacetyl-glucose, beta-D-pentaacetyl-galactose, 2-acetamido- and 3,4,6-tri-O-acetyl-2-butanamido-2-deoxy-beta-D-glucopyranosyl chlorides as well as the formation of anomeric 3-tert-butyl-4-hydroxyphenyl 4,6-di-O-acetyl-2,3-dideoxy-D-erythro-hex-2-eno-pyranosides by reaction between tert-butylhydroquinone and 3,4,6-tri-O-acetyl-D-glucal. All compounds, except 3-tert-butyl-4-hydroxyphenyl alpha- and
H Jijakli et al.
Endocrine, 10(3), 219-224 (1999-09-14)
The metabolism of alpha-D-glucose pentaacetate and its positive insulinotropic action in isolated rat pancreatic islets are both unexpectedly resistant to D-mannoheptulose, as judged from experiments conducted over 90-120 min incubation. In the present study, the possible effects of the heptose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service