Skip to Content
Merck
All Photos(1)

Documents

284998

Sigma-Aldrich

Adrenosterone

98%

Synonym(s):

11-Ketoandrostenedione, 4-Androstene-3,11,17-trione, Androst-4-ene-3,11,17-trione, Reichstein’s substance G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

optical activity

[α]20/D +300°, c = 1 in chloroform

mp

219-222 °C (lit.)

SMILES string

C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O

InChI

1S/C19H24O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h9,13-14,17H,3-8,10H2,1-2H3/t13-,14-,17+,18-,19-/m0/s1

InChI key

RZRPTBIGEANTGU-IRIMSJTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adrenosterone is a steroid hormone that is used as a dietary supplement to decrease fat and increase muscle mass.

Application

Adrenosterone has been used to study its effect on cell morphology, cell number and myogenic differentiation in killifish cell line, KFE-5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ignasi Forné et al.
Proteomics, 9(8), 2171-2181 (2009-03-27)
In the farmed flatfish Senegalese sole, F1 males reared in captivity often show lower sperm production and fertilization capacity than wild-caught males. To gain insights into the molecular mechanisms that may be altered in the F1 testis, we used 2-D
Lance Brooker et al.
Drug testing and analysis, 1(11-12), 587-595 (2010-04-01)
Adrenosterone (androst-4-ene-3,11,17-trione, 11-oxoandrostenedione) is an endogenous steroid hormone that has been promoted as a dietary supplement capable of reducing body fat and increasing muscle mass. It is proposed that adrenosterone may function as an inhibitor of the 11beta-hydroxysteroid dehydrogenase type
I Mayer et al.
Hormones and behavior, 28(2), 181-190 (1994-06-01)
The onset of spawning behavior in male salmonids can be closely correlated to circulating levels of the progestin 17 alpha,20 beta-dihydroxy-4-pregnen-3-one (20-P). However, evidence that 20-P plays a causal role in the development and maintenance of spawning behavior in male
I Mayer et al.
General and comparative endocrinology, 109(2), 223-231 (1998-02-25)
Somatolactin (SL) is a hormone recently isolated and characterized from fish pituitaries. Although the functions of SL are still largely unknown, it has been implicated in reproduction. In this study, the effects of gonadal steroids on SL secretion were investigated
René Ullrich et al.
Journal of inorganic biochemistry, 183, 84-93 (2018-04-01)
Two unspecific peroxygenases (UPO, EC 1.11.2.1) from the basidiomycetous fungi Marasmius rotula and Marasmius wettsteinii oxidized steroids with hydroxyacetyl and hydroxyl functionalities at C17 - such as cortisone, Reichstein's substance S and prednisone - via stepwise oxygenation and final fission

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service