Skip to Content
Merck
All Photos(2)

Key Documents

282979

Sigma-Aldrich

3-Ethyl-2,4-pentanedione, mixture of tautomers

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(C2H5)COCH3
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

80-81 °C/20 mmHg (lit.)

density

0.953 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCC(C(C)=O)C(C)=O

InChI

1S/C7H12O2/c1-4-7(5(2)8)6(3)9/h7H,4H2,1-3H3

InChI key

GUARKOVVHJSMRW-UHFFFAOYSA-N

Application

3-Ethyl-2,4-pentanedione has been used in the preparation of:
  • N,N,N,N-tetradentate macrocyclic ligand[1]
  • vanadyl 3-ethylacetylacetonate, its effects in non-ketotic, streptozotocin-diabetic rats has been studied[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soni Rani et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(5), 1507-1514 (2011-02-22)
Palladium(II), platinum(II), ruthenium(III) and iridium(III) complexes of general stoichiometry [PdL]Cl(2), [PtL]Cl(2), [Ru(L)Cl(2)]Cl and [Ir(L)Cl(2)]Cl are synthesized with a tetradentate macrocyclic ligand, derived from 2,6-diaminopyridine with 3-ethyl 2,4-pentanedione. Ligand was characterized on the basis of elemental analyses, IR, mass, and (1)H
B A Reul et al.
British journal of pharmacology, 126(2), 467-477 (1999-03-17)
1. Vanadium compounds can mimic actions of insulin through alternative signalling pathways. The effects of three organic vanadium compounds were studied in non-ketotic, streptozotocin-diabetic rats: vanadyl acetylacetonate (VAc), vanadyl 3-ethylacetylacetonate (VEt), and bis(maltolato)oxovanadium (VM). A simple inorganic vanadium salt, vanadyl
Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the
C Berger et al.
Human reproduction (Oxford, England), 30(4), 800-811 (2015-03-06)
Does ulipristal acetate (UPA) used for emergency contraception (EC) interfere with the human embryo implantation process? UPA, at the dosage used for EC, does not affect human embryo implantation process, in vitro. A single pre-ovulatory dose of UPA (30 mg)
Valdeir Arantes et al.
Biotechnology for biofuels, 7, 87-87 (2014-07-01)
There is still considerable debate regarding the actual mechanism by which a "cellulase mixture" deconstructs cellulosic materials, with accessibility to the substrate at the microscopic level being one of the major restrictions that limits fast, complete cellulose hydrolysis. In the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service