Skip to Content
Merck
All Photos(3)

Key Documents

278904

Sigma-Aldrich

Potassium dioxide

powder

Synonym(s):

Potassium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KO2
CAS Number:
Molecular Weight:
71.10
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

form

powder

SMILES string

[K+].[O][O-]

InChI

1S/K.HO2/c;1-2/h;1H/q+1;/p-1

InChI key

DUPSQGGNCHNYTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Hariri et al.
Scientific reports, 9(1), 11378-11378 (2019-08-08)
LED-based photoacoustic imaging has practical value in that it is affordable and rugged; however, this technology has largely been confined to anatomic imaging with limited applications into functional or molecular imaging. Here, we report molecular imaging reactive oxygen and nitrogen
Anton Misak et al.
Antioxidants (Basel, Switzerland), 9(10) (2020-10-01)
Lipid hydroperoxides play an important role in various pathophysiological processes. Therefore, a simple model for organic hydroperoxides could be helpful to monitor the biologic effects of endogenous and exogenous compounds. The electron paramagnetic resonance (EPR) spin-trapping technique is a useful
Hongtao Zhao et al.
Free radical biology & medicine, 34(11), 1359-1368 (2003-05-22)
Hydroethidine (HE) or dihydroethidium (DHE), a redox-sensitive probe, has been widely used to detect intracellular superoxide anion. It is a common assumption that the reaction between superoxide and HE results in the formation of a two-electron oxidized product, ethidium (E+)
Neeraj Chauhan et al.
Eukaryotic cell, 2(5), 1018-1024 (2003-10-14)
Ssk1p of Candida albicans is a putative response regulator protein of the Hog1 two-component signal transduction system. In Saccharomyces cerevisiae, the phosphorylation state of Ssk1p determines whether genes that promote the adaptation of cells to osmotic stress are activated. We
T Itoh et al.
Bioorganic & medicinal chemistry, 8(8), 1983-1989 (2000-09-26)
N-Aminoazaaromatics were found to react with nitric oxide in the presence of oxygen to afford deaminated products in high yields. The reaction proceeded almost instantaneously in various solvents including water, and one to two equivalent of NO was consumed depending

Articles

Oxidation and reduction reactions are some of the most common transformations encountered in organic synthesis, and are some of the organic chemist’s most powerful tools for creating novel products. Below is a list of the most commonly used oxidizing and reducing agents currently available in our catalog.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service