All Photos(1)
About This Item
Linear Formula:
(HO)2C10H5CO2H
CAS Number:
Molecular Weight:
204.18
Beilstein:
3288005
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
powder
mp
265 °C±10 °C (dec.) (lit.)
SMILES string
OC(=O)c1cc2cc(O)ccc2cc1O
InChI
1S/C11H8O4/c12-8-2-1-6-5-10(13)9(11(14)15)4-7(6)3-8/h1-5,12-13H,(H,14,15)
InChI key
QMWOUSYSNFCKAZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3,7-Dihydroxy-2-naphthoic acid served as a substrate for the enzyme NcsB1 (an O-methyltransferase involved in the biosynthesis of the enediyne antitumor antibiotic neocarzinostatin) in the NcsB1-catalyzed O-methylation of 2,7-dihydroxy-5-methyl-1-naphthoic acid.
Application
3,7-Dihydroxy-2-naphthoic acid was used in the synthesis of 3-hydroxy-7-methoxy-2-naphthoic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yinggang Luo et al.
The Journal of biological chemistry, 283(21), 14694-14702 (2008-04-05)
Neocarzinostatin, a clinical anticancer drug, is the archetypal member of the chromoprotein family of enediyne antitumor antibiotics that are composed of a nonprotein chromophore and an apoprotein. The neocarzinostatin chromophore consists of a nine-membered enediyne core, a deoxyaminosugar, and a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service