211281
9-Phenanthrol
technical grade
Synonym(s):
9-Hydroxyphenanthrene
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
mp
139-143 °C (lit.)
SMILES string
Oc1cc2ccccc2c3ccccc13
InChI
1S/C14H10O/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9,15H
InChI key
DZKIUEHLEXLYKM-UHFFFAOYSA-N
Related Categories
Application
9-Phenanthrol was used to investigate C K-edge and O K-edge near-edge X-ray absorption fine structure (NEXAFS) spectra of single-wall carbon nanotubes.
Biochem/physiol Actions
9-Phenanthrol is inhibitor of the transient receptor potential melastatin 4 (TRPM) channel, a Ca2+ -activated non-selective cation channel.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stefano Masoli et al.
Communications biology, 3(1), 222-222 (2020-05-10)
The cerebellar granule cells (GrCs) are classically described as a homogeneous neuronal population discharging regularly without adaptation. We show that GrCs in fact generate diverse response patterns to current injection and synaptic activation, ranging from adaptation to acceleration of firing.
Amy C Smith et al.
American journal of physiology. Cell physiology, 304(5), C467-C477 (2013-01-11)
Members of the transient receptor potential (TRP) channel superfamily, including the Ca(2+)-activated monovalent cation-selective TRP melastatin 4 (TRPM4) channel, have been recently identified in the urinary bladder. However, their expression and function at the level of detrusor smooth muscle (DSM)
Te-Ling Lu et al.
Biomolecules, 10(2) (2020-01-30)
: GAL-021 has recently been developed as a novel breathing control modulator. However, modifications of ionic currents produced by this agent remain uncertain, although its efficacy in suppressing the activity of big-conductance Ca2+-activated K+ (BKCa) channels has been reported. In
Ana Mrejeru et al.
The Journal of physiology, 589(Pt 10), 2497-2514 (2011-04-14)
Nigral dopamine neurons are transiently activated by high frequency glutamatergic inputs relaying reward-predicting sensory information. The tonic firing pattern of dopamine cells responds to these inputs with a transient burst of spikes that requires NMDA receptors. Here, we show that
T Grand et al.
British journal of pharmacology, 153(8), 1697-1705 (2008-02-26)
TRPM4 and TRPM5 are calcium-activated non-selective cation channels with almost identical characteristics. TRPM4 is detected in several tissues including heart, kidney, brainstem, cerebral artery and immune system whereas TRPM5 expression is more restricted. Determination of their roles in physiological processes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service