Skip to Content
Merck
All Photos(1)

Key Documents

258571

Sigma-Aldrich

2,2-Dichloropropane

98%

Synonym(s):

Isopropylidene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CCl2CH3
CAS Number:
Molecular Weight:
112.99
Beilstein:
1731150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

68-69 °C (lit.)

mp

−35 °C (lit.)

density

1.082 g/mL at 25 °C (lit.)

SMILES string

CC(C)(Cl)Cl

InChI

1S/C3H6Cl2/c1-3(2,4)5/h1-2H3

InChI key

ZEOVXNVKXIPWMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dehydrohalogenation of 2,2-dichloropropane sorbed in micropores of several model mineral solids has been investigated. Polymorphism of 2,2-dichloropropane has been investigated by thermal and X-ray powder diffraction experiments.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polymorphism of 2, 2-dichloropropane: Crystallographic characterization of the ordered and disordered phases.
Negrier P, et al.
Chemistry of Materials, 14(5), 1921-1929 (2002)
Hefa Cheng et al.
Environmental science & technology, 42(8), 2879-2885 (2008-05-24)
Nanometer scale pores are ubiquitous in porous geologic media (soils and sediments). Sorption of organic contaminants in micropores (< or = 2 nm) can inhibittheir hydrolytic transformation due to the limited availability of reactive water within hydrophobic micropore spaces. As

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service