Skip to Content
Merck
All Photos(1)

Key Documents

D72603

Sigma-Aldrich

2,3-Dichloro-1-propene

98%

Synonym(s):

2,3-Dichloro-1-propene, 2,3-Dichloropropene, 2,3-Dichloropropylene, 2-Chloroallyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CCl=CH2
CAS Number:
Molecular Weight:
110.97
Beilstein:
1361491
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.8 (vs air)

Quality Level

vapor pressure

44 mmHg ( 20 °C)

Assay

98%

form

liquid

expl. lim.

7.8 %

refractive index

n20/D 1.461 (lit.)

bp

94 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

ClCC(Cl)=C

InChI

1S/C3H4Cl2/c1-3(5)2-4/h1-2H2

InChI key

FALCMQXTWHPRIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Medinsky et al.
Toxicology letters, 23(1), 119-125 (1984-10-01)
2,3-Dichloropropene (2,3-DCP), a component of commercial fumigants and nematocides, was mixed with [14C]2,3-DCP and given to rats by peroral (p.o.) or intraperitoneal (i.p.) administration. Urine, feces, and expired air were collected over 72 h. Excretion of radioactivity in urine predominated
Roland P H Schmitz et al.
Environmental science & technology, 41(21), 7370-7375 (2007-11-30)
The reductive dehalogenation of chlorinated propenes was studied with the tetrachloroethene reductive dehalogenase purified from Sulfurospirillum multivorans to obtain indications for a radical mechanism of this reaction. When reduced methyl viologen (MV), which is a radical cation, was applied as
E Eder et al.
Drug metabolism and disposition: the biological fate of chemicals, 16(1), 60-68 (1988-01-01)
At the present time, comprehensive metabolism studies of 2,3-dichloro-1-propene (2,3-DCP) have not yet been reported. We have investigated the biotransformation of 2,3-DCP using female Wistar rats in order to elucidate the bioactivation mechanisms. 175 mg/kg, 1,3-14C-2,3-DCP in corn oil was
J S Dutcher et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(5), 997-1005 (1985-10-01)
2,3-Dichloropropene (DCP) is an intermediate used in the manufacture of carbamate herbicides and there is potential for human exposure during the manufacturing process. DCP is a known mutagen in bacteria systems and some structural analogs of DCP are carcinogenic. Since
F R Johannsen et al.
Journal of toxicology and environmental health, 33(3), 291-302 (1991-07-01)
Groups of 15 male and 15 female Sprague-Dawley rats were exposed to 1 of 3 chloropropene (2,3-Di = DCP; 1,2,3-Tri = TRCP; and 1,1,2,3-Tetra = TECP) vapors to provide information on repeated exposures and the potential for reproductive impairment by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service