Skip to Content
Merck
All Photos(3)

Key Documents

253898

Sigma-Aldrich

tert-Butyl ethyl ether

99%

Synonym(s):

2-Ethoxy-2-methylpropane, Ethyl tert-butyl ether, Ethyl tert-butyl oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC(CH3)3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731469
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021026
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

155 mmHg ( 25 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.375 (lit.)

bp

72-73 °C (lit.)

mp

−97 °C (lit.)

solubility

water: soluble 1.2 g/100g at 20 °C(lit.)

density

0.742 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCOC(C)(C)C

InChI

1S/C6H14O/c1-5-7-6(2,3)4/h5H2,1-4H3

InChI key

NUMQCACRALPSHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solid-phase microextraction of tert-butyl ethyl ether in human urine by using single-walled carbon nanotubes as adsorbent has been reported.

Application

tert-Butyl ethyl ether has been used as gasoline oxygenate and its biodegradation by propane-oxidizing bacteria has been studied.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel Müller et al.
Environmental pollution (Barking, Essex : 1987), 240, 639-646 (2018-05-18)
Microplastics are increasingly entering marine, limnic and terrestrial ecosystems worldwide, where they sorb hydrophobic organic contaminants. Here, the sorption behavior of the fuel-related water contaminants benzene, toluene, ethyl benzene and xylene (BTEX) and four tertiary butyl ethers to virgin and
Yuzo Asano et al.
Drug and chemical toxicology, 34(3), 311-317 (2011-06-09)
Ethyl tertiary-butyl ether (ETBE) is commonly used as an oxygenated gasoline additive. In this study, the prenatal developmental toxicity of ETBE was determined in rabbits. New Zealand white rabbits were given ETBE by gavage at 100, 300, or 1,000 mg/kg/day
Akihiro Hagiwara et al.
Toxicology, 289(2-3), 160-166 (2011-08-26)
The modifying potential of ethyl tertiary-butyl ether (ETBE) on tumor development was investigated in a medium-term multi-organ carcinogenesis bioassay using male F344 rats. Animals were sequentially given 5 carcinogens with different target sites in the first 4 weeks for multi-organ
Yoko Eitaki et al.
Journal of occupational health, 53(6), 423-431 (2011-10-15)
In order to measure occupational exposure concentrations of ethyl tertiary-butyl ether (ETBE), we developed a diffusive sampling method for monitoring ETBE and performed an ETBE exposure assessment. The applicability of diffusive samplers was examined by exposing the samplers to ETBE
Ann de Peyster
Birth defects research. Part B, Developmental and reproductive toxicology, 89(3), 239-263 (2010-06-15)
Ethyl t-butyl ether (ETBE) is a motor fuel oxygenate used in reformulated gasoline. Knowledge of developmental and reproductive toxicity potential of ETBE is critical for making informed decisions about acceptance and regulations. This review discusses toxicology studies providing information about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service