Skip to Content
Merck
All Photos(1)

Key Documents

95251

Supelco

Diisopropyl ether

analytical standard

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

827 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.006% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

1-21 %, 100 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.367 (lit.)
n20/D 1.368

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisopropyl ether is a secondary ether , which finds applications as an extractant and oxygenate gasoline additive.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

recommended

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Introduction to Spectroscopy (2016)
Organic Chemistry: Chemistry, Organic chemistry (2016)
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and
R van Gijn et al.
Journal of chromatography. B, Biomedical applications, 667(2), 269-276 (1995-05-19)
The staurosporine derivative, N-benzoylstaurosporine (CGP 41 251; I), is a protein kinase C inhibitor that has been selected for phase I clinical evaluation in cancer patients. We have developed a selective and sensitive assay of the drug and three potential
R Gonzalez-Olmos et al.
Chemosphere, 71(11), 2098-2105 (2008-02-27)
Methyl tert-butyl ether (MTBE) is the most widely used oxygenate in gasoline blending and has become one of the world's most widespread groundwater and surface water pollutants. Alternative oxygenates to MTBE, namely ethyl tert-butyl ether (ETBE), tert-amyl ether (TAME) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service