Skip to Content
Merck
All Photos(1)

Key Documents

248517

Sigma-Aldrich

4-Acetoxybenzoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
1871997
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Quality Level

Assay

96%

mp

190-194 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

CC(=O)Oc1ccc(cc1)C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

GDBUZIKSJGRBJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics of the bulk polymerization of 4-acetoxybenzoic acid has been investigated. Mono and submono layers of 4-acetoxybenzoic acid chemisorbed on oxidized aluminum has been studied by IR-absorption spectroscopy.

Application

4-Acetoxybenzoic acid has been used in the preparation of poly(4-hydroxybenzoate).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anomalous intensity effects in the IR spectrum of a densely packed monolayer of 4-acetoxybenzoic acid immobilized on oxidized aluminum. Conformational and vibrational analysis with AM1.
Sondag AHM, et al.
Langmuir, 8(4), 1127-1135 (1992)
Kinetics of the polymerization of 4-acetoxybenzoic acid and 6-acetoxy-2-naphthoic acid.
Williams PA, et al.
Macromolecules, 29(6), 1874-1879 (1996)
Polymer single crystals of poly (4-hydroxybenzoate). I. Morphology.
Lieser G, et al.
Journal of Polymer Science. Part B, Polymer Physics, 21(9), 1599-1609 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service