Skip to Content
Merck
All Photos(3)

Documents

248509

Sigma-Aldrich

(S)-(−)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride solution

0.1 M in methylene chloride

Synonym(s):

(S)-(−)-N-(Trifluoroacetyl)prolyl chloride solution, TPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7ClF3NO2
CAS Number:
Molecular Weight:
229.58
Beilstein:
480791
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical purity

ee: 95% (GLC)

concentration

0.1 M in methylene chloride

refractive index

n20/D 1.424

density

1.308 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)N1CCC[C@H]1C(Cl)=O

InChI

1S/C7H7ClF3NO2/c8-5(13)4-2-1-3-12(4)6(14)7(9,10)11/h4H,1-3H2/t4-/m0/s1

InChI key

NUOYJPPISCCYDH-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride can be used as a chiral derivatization reagent:
  • In the chiral separation of psychoactive, cathinone- and amphetamine-related drugs using GC-MS technique.
  • In the estimation of cathinone related drug enantiomers in biological samples like urine and plasma using GC-MS technique.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Du et al.
European review for medical and pharmacological sciences, 23(15), 6570-6578 (2019-08-06)
Papillary thyroid carcinoma (PTC) is the most common type of thyroid malignancy with physiological microRNA (miR) pathomorphological changes. MiR-548c-3p participates in multiple processes of tumor development and progression. However, the role of miR-548c-3p in PTC and the underlying mechanisms remain
Luis M Valor et al.
PloS one, 12(8), e0183264-e0183264 (2017-08-18)
Plasma cells (PC) represent the heterogeneous final stage of the B cells (BC) differentiation process. To characterize the transition of BC into PC, transcriptomes from human naïve BC were compared to those of three functionally-different subsets of human in vivo-generated
Stefan Mohr et al.
Journal of chromatography. A, 1269, 352-359 (2012-10-13)
Since cathinone derivatives gained high popularity on the recreational drugs market within the past 5 years the development of analytical methods for the achiral and chiral determination of this substance class is of great interest. Not at least because it
Simultaneous quantitative determination of synthetic cathinone enantiomers in urine and plasma using GC-NCI-MS
Alremeithi R, et al.
Journal of Analytical Methods in Chemistry, 2018 (2018)
Lorenz Hirt et al.
Journal of neuroscience research, 96(2), 194-206 (2017-04-19)
Here we assess the potential functional role of increased aquaporin 9 (APQ9) in astrocytes. Increased AQP9 expression was achieved in primary astrocyte cultures by transfection of a plasmid-containing green fluorescent protein fused to either wild-type or mutated human AQP9. Increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service