Skip to Content
Merck
All Photos(1)

Key Documents

77879

Supelco

(R)-(+)-α-Methylbenzylamine

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(R)-(+)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
2410916
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D +30±1°, c = 10% in ethanol

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.526 (lit.)
n20/D 1.528

bp

187-189 °C (lit.)

density

0.952 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1

InChI key

RQEUFEKYXDPUSK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-α-Methylbenzylamine is a high quality, useful chiral derivatization reagent for all analytical applications, specific to GC in the chiral field. It is specially selected to meet the requirements for derivatization reagents for enantiomeric excess determinations.

Application

(R)-(+)-a-Methylbenzylamine also known as (R)-(+)-1-Phenylethylamine may be used in resolution of a chiral arylalkylamine involving high-conversion enantioselective condensation with capric acid followed by hydrolysis to yield corresponding (R)-(+)-amide.

Other Notes

Chiral amine used for the determination of the enantiomeric purity of acids

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.W. Souter
Chromatographic Separations of Stereoisomers null
W.H. Pirkle and J. Finn et al.
Asymmetric Synthesis, 1 (1983)
Easy-on, easy-off?resolution of chiral 1-phenylethylamine catalyzed by Candida antarctica lipase B.
Torres-Gavilan, A., et al.
Tetrahedron Asymmetry, 18.22, 2621-2624 (2007)
Carlos Fernandes et al.
The Journal of organic chemistry, 74(8), 3217-3220 (2009-03-17)
A short, convenient, gram scale protocol has been established to allow facile access to all four stereoisomers of 2-aminocyclobutanecarboxylic acid, each in enantiomerically pure form (ee >99%). Starting from the readily available cis racemate, the procedure combines efficient alpha-phenylethylamine derivative
Paul E Harrington et al.
Current medicinal chemistry, 14(28), 3027-3034 (2008-01-29)
The calcium sensing receptor (CaR) is a G protein-coupled receptor (GPCR) that plays a fundamental role in serum calcium homeostasis. The CaR is expressed on the chief cells of the parathyroid gland and is responsible for controlling the secretion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service