Skip to Content
Merck
All Photos(1)

Key Documents

242934

Sigma-Aldrich

Ethyl iodoacetate

98%

Synonym(s):

Ethyl 2-iodoacetate, Ethyl monoiodoacetate, Iodoacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2COOC2H5
CAS Number:
Molecular Weight:
214.00
Beilstein:
741934
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.503 (lit.)

bp

179-180 °C (lit.)

density

1.808 g/mL at 25 °C (lit.)

functional group

ester
iodo

storage temp.

2-8°C

SMILES string

CCOC(=O)CI

InChI

1S/C4H7IO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

MFFXVVHUKRKXCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl iodoacetate, also known as iodoacetic acid ethyl ester, serves as an alkylating agent and a radical precursor in organic synthesis. It is also commonly used as a lachrymatory agent.

Application

Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Brucelle et al.
The Journal of organic chemistry, 78(12), 6245-6252 (2013-06-01)
The preparation of trans-2,3-disubstituted indolines from 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade using ethyl iodoacetate and triethylborane is described. Further lactamization afforded substituted benzopyrrolizidinones with excellent diastereomeric ratios. The radical cascade/lactamization sequence was efficiently applied to the synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service