Skip to Content
Merck
All Photos(3)

Key Documents

205389

Sigma-Aldrich

Hexamethyldisiloxane

≥98%

Synonym(s):

HMDSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOSi(CH3)3
CAS Number:
Molecular Weight:
162.38
Beilstein:
1736258
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.377 (lit.)

bp

101 °C (lit.)
101 °C

mp

−59 °C (lit.)

density

0.764 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethyldisiloxane (HMDSO), a linear polydisiloxane, is an organosilicon reagent commonly utilized as a source for plasma enhanced chemical vapor deposition (PE-CVD) of thin films of silicon compounds. It is also employed as a substitute to silane in silicon integrated circuit technology. Its dissociative ionization by electron impact has been studied by Fourier transform mass spectrometry. Synthesis of plasma-polymerized HMDSO thin films by atmospheric pressure glow (APG) discharge has been reported. Crystals of HMDSO at 148K are monoclinic. Addition of HMDSO to P4S10, improves its efficiency as thionating agent.

Application

Hexamethyldisiloxane can be used:
  • To fabricate a gas diffusion layer (GDL) with controlled hydrophobic silicone nano-layer by the dry deposition process.
  • To prepare silver nanoparticles coated with silica using arginine and glucose as the catalyst and the reducing agent, respectively.

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal and molecular structures of disiloxane (at 108 K) and hexamethyldisiloxane (at 148 K).
Barrow MJ, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 35(9), 2093-2099 (1979)
Synthesis of plasma-polymerized tetraethoxysilane and hexamethyldisiloxane films prepared by atmospheric pressure glow discharge.
Sawada Y, et al.
Journal of Physics D: Applied Physics, 28(8), 1661-1661 (1995)
Ion chemistries in hexamethyldisiloxane.
Jiao CQ, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 23(5), 1295-1304 (2005)
Tae-Jun Ko et al.
ACS applied materials & interfaces, 7(9), 5506-5513 (2015-02-18)
Reliable operation of a proton exchange membrane fuel cell requires proper water management to prevent water flooding in porous carbon materials such as the gas diffusion layer (GDL). In contrast to the conventional GDL that uses the "wet" dip-coating process
Israel Sifuentes-Nieves et al.
International journal of biological macromolecules, 144, 682-689 (2019-12-21)
In the present study, the effect of plasma treatment on the structural properties of three granular corn starches (normal, Hylon V and Hylon VII) was investigated. Thermal (TGA/DSC), structural (XRD/FTIR) and chemical (XPS) properties were evaluated. Plasma treatment resulted in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service