Skip to Content
Merck
All Photos(1)

Key Documents

234435

Sigma-Aldrich

(2-Chloroethyl)trimethylammonium chloride

98%

Synonym(s):

Chlormequat chloride, Chlorocholine chloride, Choline dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2Cl
CAS Number:
Molecular Weight:
158.07
Beilstein:
3563994
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

239-243 °C (dec.) (lit.)

SMILES string

[Cl-].C[N+](C)(C)CCCl

InChI

1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1

InChI key

UHZZMRAGKVHANO-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin T Sørensen et al.
International journal of andrology, 29(1), 129-133 (2006-02-10)
This paper summarizes the consequences of exposure to chlormequat, a plant growth regulator, on reproduction in mammals. Plant growth regulators are chemicals used to manipulate plant growth, flowering and fruit yield. In grain crops, plant growth regulators are applied to
M Careri et al.
Rapid communications in mass spectrometry : RCM, 16(19), 1821-1826 (2002-09-25)
A rapid method has been devised for the direct determination of chlormequat in tomato samples. No clean-up is required, and analysis uses ion-exchange liquid chromatography/tandem mass spectrometry interfaced with electrospray ionization (LC/ESI-MS/MS). A cation-exchange column was used with an aqueous
Xing-li Guo et al.
Ecotoxicology and environmental safety, 73(4), 642-646 (2010-01-12)
A simple and reliable analytical method for chlormequat residues in cotton and soil was established in this study. The residual levels and dissipation rates of chlormequat in cotton crops and soil were determined by High Performance Liquid Chromatography-Mass Spectroscopy (HPLC-MS).
Oscar Núñez et al.
Journal of mass spectrometry : JMS, 39(8), 873-883 (2004-08-27)
Fragmentation studies using both an ion-trap mass analyzer and a hybrid quadrupole time-of-flight (Q-TOF) mass spectrometer were performed in order to establish the fragmentation pathways of organic molecules. A general strategy combining MSn data (n = 1-4) in an ion-trap
René K Juhler et al.
Environmental science and pollution research international, 17(6), 1245-1256 (2010-02-24)
Chlormequat (Cq) is a plant growth regulator used throughout the world. Despite indications of possible effects of Cq on mammalian health and fertility, little is known about its fate and transport in subsurface environments. The aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service