Skip to Content
Merck
All Photos(5)

Key Documents

154393

Sigma-Aldrich

4-Acetylbenzonitrile

greener alternative

99%

Synonym(s):

4′-Cyanoacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4CN
CAS Number:
Molecular Weight:
145.16
Beilstein:
1932887
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

greener alternative product score

old score: 18
new score: 3
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

56-59 °C (lit.)

functional group

ketone
nitrile

greener alternative category

SMILES string

CC(=O)c1ccc(cc1)C#N

InChI

1S/C9H7NO/c1-7(11)9-4-2-8(6-10)3-5-9/h2-5H,1H3

InChI key

NLPHXWGWBKZSJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The relaxation-assisted two-dimensional infrared (RA 2DIR) spectroscopy of 4-acetylbenzonitrile was analyzed.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom economy”, “Design for Energy Efficiency”, “Use of Renewable Feedstock” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

4-Acetylbenzonitrile was used in the synthesis of imidazole derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry V Kurochkin et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(36), 14209-14214 (2007-06-15)
A method of two-dimensional infrared (2D IR) spectroscopy called relaxation-assisted 2D IR (RA 2DIR) is proposed that utilizes vibrational energy relaxation transport in molecules to enhance cross-peak amplitudes. This method substantially increases the range of distances accessible by 2D IR
Sham M Sondhi et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 4(2), 146-154 (2008-03-14)
A series of thiophene derivatives 1a-d & 2a-c were synthesized by condensation of 5-nitro-2-thiophene carboxaldehyde with mono and diamines respectively. Various imidazole derivatives 3a-c were obtained by condensing 4-(2-ethylamino)-1H-imidazole with 4-acetylpyridine, 2-acetylpyridine and 4-acetylbenzonitrile respectively. Pyridine derivatives 4a-e were synthesized
Koji Uwai et al.
Bioorganic & medicinal chemistry, 16(3), 1084-1089 (2007-11-17)
Stereoselective reductive metabolism of various p-substituted acetophenone derivatives was studied using isolated rat liver 3alpha-hydroxysteroid dehydrogenase (3alpha-HSD). Kinetic experiments were performed and analyzed by measuring the products by HPLC using a chiral column. The results demonstrated that the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service