Skip to Content
Merck
All Photos(2)

Documents

228583

Sigma-Aldrich

3-Chloro-4-fluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)NH2
CAS Number:
Molecular Weight:
145.56
Beilstein:
1562786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

227-228 °C (lit.)

mp

42-44 °C (lit.)

SMILES string

Nc1ccc(F)c(Cl)c1

InChI

1S/C6H5ClFN/c7-5-3-4(9)1-2-6(5)8/h1-3H,9H2

InChI key

YSEMCVGMNUUNRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The exposure of workers to 3-chloro-4-fluoroaniline (CFA) was monitored by an HPLC method.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P J Boogaard et al.
Environmental health perspectives, 102 Suppl 6, 23-25 (1994-10-01)
In two studies, involving 75 and 72 workers, potential exposure to 3-chloro-4-fluoroaniline (CFA) was biologically monitored by determination of its main urinary metabolite 2-amino-4-chloro-5-fluorophenol sulfate (CFA-S). As this method only allows the detection of recent exposure, analysis of CFA adducts
C V Eadsforth et al.
Journal of analytical toxicology, 12(6), 330-333 (1988-11-01)
An HPLC method for monitoring exposure of workers to 3-chloro-4-fluoroaniline (CFA) is described. It is based on the detection of a major urinary metabolite, 2-amino-4-chloro-5-fluorophenyl sulphate, and is superior to the previously adopted GC method. The limit of detection for
M K Baldwin et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(2), 135-144 (1980-02-01)
1. 3-Chloro-4-fluorol[14C]aniline, orally administered to a female dog (0.135 mg/kg), was eliminated in the urine as 2-amino-4-chloro-5-fluorophenyl sulphate (83% in 48 h). 2. When 3-chloro-4-fluoro[14C]aniline was administered orally to male rats (ca 2.3 mg/kg), the 0-24 h urine contained 2-amino-4-chloro-5-fluorophenyl
C J Duckett et al.
Xenobiotica; the fate of foreign compounds in biological systems, 36(1), 59-77 (2006-03-02)
The metabolic fate of 3-chloro-4-fluoroaniline was investigated in rat following intraperitoneal (i.p.) administration at 5 and 50 mg kg(-1) using a combination of HPLC-MS, HPLC-MS/MS, (19)F-NMR spectroscopy, HPLC-NMR spectroscopy and high-pressure liquid chromatography-inductively coupled plasma mass spectrometry (HPLC-ICPMS) with (35)Cl
Vasili Travkin et al.
FEMS microbiology letters, 209(2), 307-312 (2002-05-15)
In this paper we report the isolation and characterization of an anaerobic enrichment culture as well as of a Rhodococcus sp. strain 2 capable of degrading 3,4-dihaloanilines under nitrate reducing conditions. Using mass spectrometry several of the intermediates formed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service