Skip to Content
Merck
All Photos(1)

Documents

D101400

Sigma-Aldrich

2,4-Difluoroaniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NH2
CAS Number:
Molecular Weight:
129.11
Beilstein:
2802556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.506 (lit.)

bp

170 °C/753 mmHg (lit.)

mp

−7.5 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(F)cc1F

InChI

1S/C6H5F2N/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

CEPCPXLLFXPZGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Chih Huang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 176-179 (2012-04-07)
We applied the two-color resonant two-photon ionization and mass-analyzed threshold ionization techniques to record the vibronic and cation spectra of 2,4-difluoroaniline. The cation spectra were recorded by ionizing via the 0(0), X(1), 6b(1), and 1(1) levels of the electronically excited
P J Boogaard et al.
Environmental health perspectives, 102 Suppl 6, 27-29 (1994-10-01)
Exposure to 2,4-difluoroaniline (DFA) was monitored by GC-MS of DFA adducts bound to hemoglobin (Hb). In two studies, involving 20 and 16 workers potentially exposed to low concentrations of DFA, median concentrations of 10 (range 1-83) and 20 (range 4-322)
Junyu Guo et al.
Environmental pollution (Barking, Essex : 1987), 225, 175-183 (2017-04-04)
Trifluoroacetic acid (TFA) in the atmosphere is produced by degradation of hydrochlorofluorocarbons and hydrofluorocarbons. In recent years, TFA has attracted global attention because of increased environmental concentrations, biological toxicity and accumulation in aqueous environments. This study focused on the mechanisms
Sergey E Korchak et al.
The Journal of chemical physics, 137(9), 094503-094503 (2012-09-11)
Effects of spin-spin interactions on the nuclear magnetic relaxation dispersion (NMRD) of protons were studied in a situation where spin ½ hetero-nuclei are present in the molecule. As in earlier works [K. L. Ivanov, A. V. Yurkovskaya, and H.-M. Vieth
Duoli Guo et al.
AAPS PharmSciTech, 13(2), 661-673 (2012-05-04)
A stability-indicating high-performance liquid chromatography method to quantify 2-(2,4-difluorophenyl)-4,5,6,7-tetrafluoroisoindoline-1,3-dione (NSC-726796) and its three main degradation products was developed. This method was used to investigate its degradation kinetics and mechanism. The reaction follows first-order kinetics and appears to be base catalyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service