All Photos(1)
About This Item
Linear Formula:
CH3C6H2(CH2OH)2OH
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
5.8 (vs air)
Assay
95%
impurities
<3% sodium hydroxide
mp
128-130 °C (lit.)
functional group
hydroxyl
SMILES string
Cc1cc(CO)c(O)c(CO)c1
InChI
1S/C9H12O3/c1-6-2-7(4-10)9(12)8(3-6)5-11/h2-3,10-12H,4-5H2,1H3
InChI key
KUMMBDBTERQYCG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2,6-Bis(hydroxymethyl)-p-cresol was used as a bridging ligand in binuclear oxovanadium(V) compound, which on exposure to white light and aerial oxygen forms an oligomeric compound.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zehong Xiang et al.
Biomaterials science, 8(21), 6025-6036 (2020-10-01)
Thrombotic and inflammatory complications induced by vascular implants remain a challenge to treat cardiovascular disease due to the lack of self-adaption and functional integrity of implants. Inspired by the dynamic remodeling of the extracellular matrix (ECM), we constructed a bio-mimic
Pabitra Baran Chatterjee et al.
Journal of the American Chemical Society, 132(45), 15842-15845 (2010-10-23)
An unprecedented single crystal-to-single crystal transformation occurs when a binuclear oxovanadium(V) compound [V(V)(2)O(2)(L)(2)] 1 involving 2,6-bis(hydroxymethyl)-p-cresol (H(3)L) as a bridging ligand is exposed simultaneously to white light and aerial oxygen to generate an oligomeric compound [V(IV)(2)O(2)(L*)(2)] 2 (H(2)L* is 3-hydroxymethyl-5-methylsalicylaldehyde).
Youngkeun Lee et al.
The Analyst, 143(8), 1780-1785 (2018-01-31)
The development of artificial peroxidases has attracted great interest because of their applications in various fields such as the chemical industry and biosensing. In this study, 2,6-bis[(bis(2-pyridylmethyl)amino)-methyl]-4-methylphenol (H-bpmp) complexes with various transition metal ions have been investigated as artificial peroxidases.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service