Skip to Content
Merck
All Photos(1)

Key Documents

219975

Sigma-Aldrich

5-Hydroxy-1-tetralone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
2437410
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

206-209 °C (lit.)

functional group

ketone

SMILES string

Oc1cccc2C(=O)CCCc12

InChI

1S/C10H10O2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1,3,5,11H,2,4,6H2

InChI key

YPPZCRZRQHFRBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Hydroxy-1-tetralone was used:
  • as internal standard during HPLC determination of 4-hydroxymephenytoin (4-OH-M) in human urine
  • as fluorescent labeling reagent during microdetection of glycosphingolipid on TLC plates
  • in synthesis of 1,2,3,4-tetrahydro-5H-1-benzazepine-quinone derivatives
  • in synthesis of new chiral oxathiane

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new chiral oxathiane: synthesis, resolution and absolute configuration determination by vibrational circular dichroism.
Solladie-Cavallo A, et al.
Tetrahedron Asymmetry, 12(18), 2605-2611 (2001)
Studies on Quinones. Part 22.'Synthesis of 1-Benzazepine-6, 9-Quinone Derivatives.
Valderrama JA, et al.
Synthetic Communications, 22(4), 629-640 (1992)
K Watanabe et al.
Journal of lipid research, 36(8), 1848-1855 (1995-08-01)
A microdetection system for glycosphingolipid analysis has been developed using 5-hydroxy-1-tetralone as the fluorescent labeling reagent. The reagents in H2SO4 permit the fluorometric detection of acidic and neutral glycosphingolipids both in test tube and on thin-layer chromatographic plates. Glycosphingolipids can
M Tanaka et al.
Journal of chromatography. B, Biomedical applications, 676(1), 87-94 (1996-02-09)
A simple and selective HPLC method for the determination of 4-hydroxymephenytoin (4-OH-M) in human urine, using a controlled potential coulometric detector equipped with a dual working electrode cell of fully porous graphite, has been developed. After acid hydrolysis of urine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service