Skip to Content
Merck
All Photos(2)

Documents

226866

Sigma-Aldrich

Silver(I) fluoride

99%

Synonym(s):

Argentous fluoride, Fluorosilver, Silver monofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgF
CAS Number:
Molecular Weight:
126.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

bp

1159 °C/1 atm (lit.)

mp

≥300 °C (lit.)

density

5.852 g/mL at 25 °C (lit.)

SMILES string

F[Ag]

InChI

1S/Ag.FH/h;1H/q+1;/p-1

InChI key

REYHXKZHIMGNSE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Silver(I)fluoride can be used as a reagent:
  • To synthesize aliphatic fluorinated products via oxidative C-H fluorination reaction using manganese-based catalysts.
  • In fluorinates homocoupling reaction of gem-difluoroalkenes.
It can also be used as a silver precursor salt to prepare silver nanoparticles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing Gao et al.
Organic letters, 16(1), 102-105 (2013-12-04)
A novel silver(I)-fluoride-mediated homocoupling reaction of β,β-difluorostyrene derivatives is described. The transformation is initiated via nucleophilic addition of silver(I) fluoride to β,β-difluorostyrenes, which is followed by dimerization of the corresponding benzylsilver intermediates. The reaction shows good substrate scope, functional group

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service